Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37798982-37799233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYBB
Entrez Gene ID
1536
Gene Name
cytochrome b-245 beta chain
Gene Aliases
AMCBX2, CGD, GP91-1, GP91-PHOX, GP91PHOX, IMD34, NOX2, p91-PHOX
UniGene
Hs.292356
Gene Chromosome Location
Chr.X: 37780017-37813461 on Build GRCh38
Transcripts
NM_000397.3

SNP Detail

SNP ID Chromosome Location
rs34065977 Chr.X: 37799136-37799136 on Build GRCh38
rs191609950 Chr.X: 37799177-37799177 on Build GRCh38
rs974905008 Chr.X: 37799117-37799117 on Build GRCh38
rs781795599 Chr.X: 37799104-37799104 on Build GRCh38
rs913681788 Chr.X: 37799196-37799196 on Build GRCh38
rs782137081 Chr.X: 37799097-37799097 on Build GRCh38
rs781962656 Chr.X: 37799139-37799139 on Build GRCh38
rs5964125 Chr.X: 37799202-37799202 on Build GRCh38
rs190924506 Chr.X: 37799024-37799024 on Build GRCh38
rs886041194 Chr.X: 37799022-37799023 on Build GRCh38
rs186579421 Chr.X: 37799161-37799161 on Build GRCh38
rs782700208 Chr.X: 37799098-37799098 on Build GRCh38
rs782756433 Chr.X: 37799016-37799016 on Build GRCh38
rs137854589 Chr.X: 37799011-37799011 on Build GRCh38
rs183050509 Chr.X: 37799130-37799130 on Build GRCh38
rs782807990 Chr.X: 37799092-37799093 on Build GRCh38

View Full Product Details