Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45650220-45650487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST1
Entrez Gene ID
8534
Gene Name
carbohydrate sulfotransferase 1
Gene Aliases
C6ST, GST-1, KS6ST, KSGal6ST, KSST
UniGene
Hs.104576
Gene Chromosome Location
Chr.11: 45647689-45665656 on Build GRCh38
Transcripts
NM_003654.5 XM_006718356.3 XM_017018459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246029 c.530A>G p.E177G Chr.11: 45650394-45650394 on Build GRCh38
6068776 c.567C>A p.N189K Chr.11: 45650357-45650357 on Build GRCh38
6628818 c.625C>T p.R209C Chr.11: 45650299-45650299 on Build GRCh38
4518054 c.507_508GG>AA p.G170R Chr.11: 45650416-45650417 on Build GRCh38
7361265 c.463G>T p.G155W Chr.11: 45650461-45650461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755387798 Chr.11: 45650410-45650410 on Build GRCh38
rs898108901 Chr.11: 45650291-45650291 on Build GRCh38
rs946310136 Chr.11: 45650449-45650449 on Build GRCh38
rs770842358 Chr.11: 45650339-45650339 on Build GRCh38
rs966526324 Chr.11: 45650390-45650390 on Build GRCh38
rs770736998 Chr.11: 45650465-45650465 on Build GRCh38
rs747607145 Chr.11: 45650416-45650416 on Build GRCh38
rs990417736 Chr.11: 45650375-45650375 on Build GRCh38
rs559545692 Chr.11: 45650426-45650426 on Build GRCh38
rs753820223 Chr.11: 45650345-45650345 on Build GRCh38
rs957906360 Chr.11: 45650357-45650357 on Build GRCh38
rs147129669 Chr.11: 45650350-45650350 on Build GRCh38
rs780753667 Chr.11: 45650331-45650331 on Build GRCh38
rs755751075 Chr.11: 45650284-45650284 on Build GRCh38
rs780463598 Chr.11: 45650396-45650396 on Build GRCh38
rs1027886195 Chr.11: 45650300-45650300 on Build GRCh38
rs1017694095 Chr.11: 45650367-45650367 on Build GRCh38
rs752079319 Chr.11: 45650402-45650402 on Build GRCh38
rs764256918 Chr.11: 45650342-45650342 on Build GRCh38
rs139471113 Chr.11: 45650351-45650351 on Build GRCh38
rs774315452 Chr.11: 45650340-45650340 on Build GRCh38
rs370921522 Chr.11: 45650386-45650386 on Build GRCh38
rs373748067 Chr.11: 45650459-45650459 on Build GRCh38
rs369492359 Chr.11: 45650413-45650413 on Build GRCh38
rs752313786 Chr.11: 45650278-45650278 on Build GRCh38
rs557691037 Chr.11: 45650255-45650255 on Build GRCh38
rs200869113 Chr.11: 45650365-45650365 on Build GRCh38
rs765383165 Chr.11: 45650358-45650358 on Build GRCh38
rs762830741 Chr.11: 45650338-45650338 on Build GRCh38
rs773156435 Chr.11: 45650337-45650337 on Build GRCh38
rs777567607 Chr.11: 45650445-45650445 on Build GRCh38
rs768176753 Chr.11: 45650315-45650315 on Build GRCh38
rs747837712 Chr.11: 45650332-45650332 on Build GRCh38
rs750719004 Chr.11: 45650256-45650256 on Build GRCh38
rs780762413 Chr.11: 45650414-45650414 on Build GRCh38
rs746564532 Chr.11: 45650296-45650296 on Build GRCh38
rs760717126 Chr.11: 45650341-45650341 on Build GRCh38
rs769631706 Chr.11: 45650417-45650417 on Build GRCh38
rs757422338 Chr.11: 45650246-45650246 on Build GRCh38

View Full Product Details