Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18809691-18809940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf78
Entrez Gene ID
100128496
Gene Name
chromosome 20 open reading frame 78
Gene Aliases
dJ1068E13.1
UniGene
Hs.664484
Gene Chromosome Location
Chr.20: 18809351-18830203 on Build GRCh38
Transcripts
NM_001242671.2 NM_001302818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6539366 c.366T>C p.G122G Chr.20: 18809866-18809866 on Build GRCh38
5013567 c.334A>G p.N112D Chr.20: 18809898-18809898 on Build GRCh38
4985694 c.334A>C p.N112H Chr.20: 18809898-18809898 on Build GRCh38
6467553 c.310C>A p.Q104K Chr.20: 18809922-18809922 on Build GRCh38
4546231 c.395G>A p.G132D Chr.20: 18809837-18809837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049588592 Chr.20: 18809895-18809895 on Build GRCh38
rs1054088884 Chr.20: 18809734-18809734 on Build GRCh38
rs972753360 Chr.20: 18809873-18809873 on Build GRCh38
rs891435807 Chr.20: 18809920-18809920 on Build GRCh38
rs539586600 Chr.20: 18809857-18809857 on Build GRCh38
rs267605850 Chr.20: 18809855-18809855 on Build GRCh38
rs577313542 Chr.20: 18809784-18809784 on Build GRCh38
rs759858998 Chr.20: 18809765-18809765 on Build GRCh38
rs565327052 Chr.20: 18809767-18809767 on Build GRCh38
rs911051129 Chr.20: 18809911-18809911 on Build GRCh38
rs748917986 Chr.20: 18809898-18809898 on Build GRCh38
rs971505694 Chr.20: 18809756-18809756 on Build GRCh38
rs757042583 Chr.20: 18809778-18809778 on Build GRCh38
rs1014961061 Chr.20: 18809918-18809918 on Build GRCh38
rs974251254 Chr.20: 18809802-18809802 on Build GRCh38
rs944074295 Chr.20: 18809838-18809838 on Build GRCh38
rs866004212 Chr.20: 18809874-18809874 on Build GRCh38
rs981129240 Chr.20: 18809757-18809757 on Build GRCh38
rs764118953 Chr.20: 18809809-18809809 on Build GRCh38
rs537731947 Chr.20: 18809909-18809909 on Build GRCh38
rs145539699 Chr.20: 18809859-18809859 on Build GRCh38
rs1023176560 Chr.20: 18809888-18809888 on Build GRCh38
rs768081306 Chr.20: 18809913-18809913 on Build GRCh38
rs1042359234 Chr.20: 18809846-18809846 on Build GRCh38
rs1054283980 Chr.20: 18809858-18809858 on Build GRCh38
rs912639361 Chr.20: 18809828-18809828 on Build GRCh38
rs773931327 Chr.20: 18809915-18809915 on Build GRCh38
rs758431698 Chr.20: 18809796-18809796 on Build GRCh38
rs765468487 Chr.20: 18809843-18809843 on Build GRCh38
rs935731439 Chr.20: 18809912-18809912 on Build GRCh38
rs566133535 Chr.20: 18809849-18809849 on Build GRCh38
rs1022700744 Chr.20: 18809754-18809754 on Build GRCh38
rs546412861 Chr.20: 18809844-18809844 on Build GRCh38
rs932785937 Chr.20: 18809810-18809810 on Build GRCh38
rs371614082 Chr.20: 18809856-18809856 on Build GRCh38
rs909856924 Chr.20: 18809852-18809852 on Build GRCh38
rs141000022 Chr.20: 18809917-18809917 on Build GRCh38
rs528229277 Chr.20: 18809812-18809812 on Build GRCh38
rs750184618 Chr.20: 18809727-18809727 on Build GRCh38
rs1000394007 Chr.20: 18809728-18809728 on Build GRCh38
rs146528664 Chr.20: 18809887-18809887 on Build GRCh38
rs757348730 Chr.20: 18809771-18809771 on Build GRCh38

View Full Product Details