Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3045509-3045764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNRH2
Entrez Gene ID
2797
Gene Name
gonadotropin releasing hormone 2
Gene Aliases
GnRH-II, LH-RHII
UniGene
Hs.129715
Gene Chromosome Location
Chr.20: 3039061-3045896 on Build GRCh38
Transcripts
NM_001310220.1 NM_001501.1 NM_178331.1 NM_178332.1 XM_017027823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687512 c.310C>A p.R104S Chr.20: 3045704-3045704 on Build GRCh38
4846036 c.314C>T p.P105L Chr.20: 3045708-3045708 on Build GRCh38
2888581 c.326C>T p.S109F Chr.20: 3045720-3045720 on Build GRCh38
4381305 c.306_307insGCCCC p.P103fs*>13 Chr.20: 3045700-3045701 on Build GRCh38
5445843 c.323_324insCCGCC p.S109fs*>7 Chr.20: 3045717-3045718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755035669 Chr.20: 3045662-3045662 on Build GRCh38
rs764704537 Chr.20: 3045649-3045649 on Build GRCh38
rs750296533 Chr.20: 3045729-3045729 on Build GRCh38
rs374040208 Chr.20: 3045661-3045661 on Build GRCh38
rs750290469 Chr.20: 3045686-3045687 on Build GRCh38
rs763863070 Chr.20: 3045704-3045705 on Build GRCh38
rs756075006 Chr.20: 3045739-3045739 on Build GRCh38
rs958702412 Chr.20: 3045583-3045583 on Build GRCh38
rs778925098 Chr.20: 3045663-3045663 on Build GRCh38
rs776242208 Chr.20: 3045641-3045641 on Build GRCh38
rs16996832 Chr.20: 3045699-3045700 on Build GRCh38
rs962175576 Chr.20: 3045535-3045535 on Build GRCh38
rs539981612 Chr.20: 3045650-3045650 on Build GRCh38
rs890149986 Chr.20: 3045632-3045632 on Build GRCh38
rs562841671 Chr.20: 3045710-3045710 on Build GRCh38
rs1043247646 Chr.20: 3045568-3045568 on Build GRCh38
rs923060898 Chr.20: 3045688-3045688 on Build GRCh38
rs774871132 Chr.20: 3045691-3045691 on Build GRCh38
rs767316458 Chr.20: 3045726-3045726 on Build GRCh38
rs767016613 Chr.20: 3045640-3045640 on Build GRCh38
rs968015569 Chr.20: 3045614-3045614 on Build GRCh38
rs67749149 Chr.20: 3045700-3045704 on Build GRCh38
rs887428093 Chr.20: 3045728-3045728 on Build GRCh38
rs758686635 Chr.20: 3045714-3045718 on Build GRCh38
rs370558288 Chr.20: 3045555-3045556 on Build GRCh38
rs574167336 Chr.20: 3045686-3045686 on Build GRCh38
rs747409278 Chr.20: 3045670-3045670 on Build GRCh38
rs781073713 Chr.20: 3045659-3045659 on Build GRCh38
rs748335284 Chr.20: 3045667-3045667 on Build GRCh38
rs761002640 Chr.20: 3045544-3045544 on Build GRCh38
rs752451333 Chr.20: 3045705-3045706 on Build GRCh38
rs763681690 Chr.20: 3045702-3045702 on Build GRCh38
rs539437304 Chr.20: 3045606-3045606 on Build GRCh38
rs16988234 Chr.20: 3045608-3045608 on Build GRCh38
rs762613660 Chr.20: 3045698-3045698 on Build GRCh38
rs544846287 Chr.20: 3045699-3045699 on Build GRCh38
rs763885965 Chr.20: 3045655-3045655 on Build GRCh38
rs535709446 Chr.20: 3045582-3045582 on Build GRCh38
rs761550854 Chr.20: 3045708-3045708 on Build GRCh38
rs1002977246 Chr.20: 3045541-3045541 on Build GRCh38
rs371531736 Chr.20: 3045642-3045642 on Build GRCh38
rs751373487 Chr.20: 3045656-3045656 on Build GRCh38
rs751246947 Chr.20: 3045704-3045704 on Build GRCh38
rs757142575 Chr.20: 3045657-3045657 on Build GRCh38
rs924533992 Chr.20: 3045592-3045592 on Build GRCh38
rs377526911 Chr.20: 3045548-3045548 on Build GRCh38
rs6138994 Chr.20: 3045592-3045592 on Build GRCh38
rs778150821 Chr.20: 3045668-3045668 on Build GRCh38
rs758057143 Chr.20: 3045653-3045653 on Build GRCh38
rs1006487686 Chr.20: 3045737-3045737 on Build GRCh38
rs747462203 Chr.20: 3045720-3045721 on Build GRCh38
rs777257234 Chr.20: 3045672-3045672 on Build GRCh38
rs778288172 Chr.20: 3045718-3045719 on Build GRCh38
rs776856452 Chr.20: 3045595-3045595 on Build GRCh38

View Full Product Details