Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138286422-138286693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25C
Entrez Gene ID
995
Gene Name
cell division cycle 25C
Gene Aliases
CDC25, PPP1R60
UniGene
Hs.656
Gene Chromosome Location
Chr.5: 138285265-138338355 on Build GRCh38
Transcripts
NM_001287582.1 NM_001287583.1 NM_001318098.1 NM_001790.4 NM_022809.3 XM_005272145.3 XM_006714739.3 XM_011543759.2 XM_011543760.2 XM_011543761.2 XM_011543762.1 XM_011543763.1 XM_011543764.1 XM_017010101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5060644 c.1099A>C p.T367P Chr.5: 138286558-138286558 on Build GRCh38
4391268 c.1054G>C p.E352Q Chr.5: 138286603-138286603 on Build GRCh38
1671897 c.1027G>T p.G343* Chr.5: 138286630-138286630 on Build GRCh38
1061289 c.1120G>A p.V374M Chr.5: 138286537-138286537 on Build GRCh38
1061287 c.1132G>T p.E378* Chr.5: 138286525-138286525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368247596 Chr.5: 138286540-138286540 on Build GRCh38
rs869074647 Chr.5: 138286595-138286596 on Build GRCh38
rs748364070 Chr.5: 138286476-138286476 on Build GRCh38
rs778271287 Chr.5: 138286573-138286573 on Build GRCh38
rs144706977 Chr.5: 138286538-138286538 on Build GRCh38
rs777727419 Chr.5: 138286529-138286529 on Build GRCh38
rs371708349 Chr.5: 138286583-138286583 on Build GRCh38
rs770854470 Chr.5: 138286484-138286484 on Build GRCh38
rs869027549 Chr.5: 138286506-138286507 on Build GRCh38
rs367857039 Chr.5: 138286650-138286650 on Build GRCh38
rs202170372 Chr.5: 138286479-138286479 on Build GRCh38
rs758553279 Chr.5: 138286556-138286556 on Build GRCh38
rs747974535 Chr.5: 138286659-138286666 on Build GRCh38
rs780632652 Chr.5: 138286448-138286448 on Build GRCh38
rs755330774 Chr.5: 138286550-138286550 on Build GRCh38
rs547482356 Chr.5: 138286449-138286449 on Build GRCh38
rs745766399 Chr.5: 138286581-138286581 on Build GRCh38
rs747954939 Chr.5: 138286473-138286473 on Build GRCh38
rs761374740 Chr.5: 138286626-138286626 on Build GRCh38
rs374330012 Chr.5: 138286491-138286491 on Build GRCh38
rs774051649 Chr.5: 138286614-138286614 on Build GRCh38
rs779620814 Chr.5: 138286500-138286501 on Build GRCh38
rs749078522 Chr.5: 138286649-138286649 on Build GRCh38
rs149379419 Chr.5: 138286537-138286537 on Build GRCh38
rs770805761 Chr.5: 138286601-138286601 on Build GRCh38
rs557788948 Chr.5: 138286525-138286525 on Build GRCh38
rs777029459 Chr.5: 138286633-138286633 on Build GRCh38
rs781019622 Chr.5: 138286576-138286576 on Build GRCh38
rs375992598 Chr.5: 138286594-138286594 on Build GRCh38
rs900866274 Chr.5: 138286527-138286527 on Build GRCh38
rs778788394 Chr.5: 138286652-138286654 on Build GRCh38
rs752875978 Chr.5: 138286554-138286554 on Build GRCh38
rs200759602 Chr.5: 138286500-138286500 on Build GRCh38
rs754687956 Chr.5: 138286649-138286652 on Build GRCh38
rs190605129 Chr.5: 138286501-138286501 on Build GRCh38
rs767096637 Chr.5: 138286627-138286627 on Build GRCh38
rs147251232 Chr.5: 138286512-138286512 on Build GRCh38
rs142916882 Chr.5: 138286570-138286570 on Build GRCh38
rs763697905 Chr.5: 138286505-138286505 on Build GRCh38
rs148571728 Chr.5: 138286568-138286568 on Build GRCh38
rs115524830 Chr.5: 138286524-138286524 on Build GRCh38
rs745420668 Chr.5: 138286667-138286667 on Build GRCh38
rs773993092 Chr.5: 138286510-138286510 on Build GRCh38
rs746715217 Chr.5: 138286553-138286553 on Build GRCh38
rs772162358 Chr.5: 138286477-138286477 on Build GRCh38
rs775024783 Chr.5: 138286503-138286503 on Build GRCh38
rs570467576 Chr.5: 138286571-138286571 on Build GRCh38
rs757236808 Chr.5: 138286575-138286575 on Build GRCh38
rs768409329 Chr.5: 138286609-138286609 on Build GRCh38
rs749015472 Chr.5: 138286598-138286598 on Build GRCh38
rs570293812 Chr.5: 138286504-138286504 on Build GRCh38
rs755748798 Chr.5: 138286501-138286501 on Build GRCh38
rs201570266 Chr.5: 138286468-138286468 on Build GRCh38

View Full Product Details