Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11506207-11506456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECSIT
Entrez Gene ID
51295
Gene Name
ECSIT signalling integrator
Gene Aliases
SITPEC
UniGene
Hs.515146
Gene Chromosome Location
Chr.19: 11505916-11529172 on Build GRCh38
Transcripts
NM_001142464.2 NM_001142465.2 NM_001243204.1 NM_016581.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
990942 c.1084G>A p.A362T Chr.19: 11506396-11506396 on Build GRCh38
7080895 c.1179C>T p.L393L Chr.19: 11506301-11506301 on Build GRCh38
6855443 c.1178T>A p.L393H Chr.19: 11506302-11506302 on Build GRCh38
4845117 c.1107G>A p.T369T Chr.19: 11506373-11506373 on Build GRCh38
7221395 c.1216G>A p.G406R Chr.19: 11506264-11506264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746478913 Chr.19: 11506397-11506397 on Build GRCh38
rs774762405 Chr.19: 11506405-11506405 on Build GRCh38
rs1045837 Chr.19: 11506233-11506233 on Build GRCh38
rs2302971 Chr.19: 11506264-11506264 on Build GRCh38
rs767782643 Chr.19: 11506354-11506354 on Build GRCh38
rs1005347301 Chr.19: 11506243-11506243 on Build GRCh38
rs767582209 Chr.19: 11506289-11506289 on Build GRCh38
rs773799891 Chr.19: 11506337-11506337 on Build GRCh38
rs746241148 Chr.19: 11506409-11506409 on Build GRCh38
rs751820230 Chr.19: 11506285-11506285 on Build GRCh38
rs778626825 Chr.19: 11506313-11506313 on Build GRCh38
rs774617724 Chr.19: 11506348-11506348 on Build GRCh38
rs753540238 Chr.19: 11506302-11506302 on Build GRCh38
rs752817300 Chr.19: 11506295-11506295 on Build GRCh38
rs765079470 Chr.19: 11506380-11506380 on Build GRCh38
rs746532327 Chr.19: 11506326-11506326 on Build GRCh38
rs556346750 Chr.19: 11506247-11506247 on Build GRCh38
rs371383315 Chr.19: 11506401-11506401 on Build GRCh38
rs766444041 Chr.19: 11506277-11506277 on Build GRCh38
rs771304747 Chr.19: 11506346-11506346 on Build GRCh38
rs771888221 Chr.19: 11506319-11506319 on Build GRCh38
rs200298350 Chr.19: 11506251-11506251 on Build GRCh38
rs750245223 Chr.19: 11506382-11506382 on Build GRCh38
rs145055878 Chr.19: 11506379-11506379 on Build GRCh38
rs772211646 Chr.19: 11506425-11506425 on Build GRCh38
rs752854709 Chr.19: 11506346-11506347 on Build GRCh38
rs759797788 Chr.19: 11506352-11506352 on Build GRCh38
rs753796849 Chr.19: 11506374-11506374 on Build GRCh38
rs759735168 Chr.19: 11506286-11506286 on Build GRCh38
rs770359160 Chr.19: 11506336-11506336 on Build GRCh38
rs756026923 Chr.19: 11506296-11506296 on Build GRCh38
rs757084368 Chr.19: 11506305-11506305 on Build GRCh38
rs758068826 Chr.19: 11506393-11506393 on Build GRCh38
rs773661666 Chr.19: 11506274-11506274 on Build GRCh38
rs138790462 Chr.19: 11506403-11506403 on Build GRCh38
rs777713525 Chr.19: 11506298-11506298 on Build GRCh38
rs760745453 Chr.19: 11506359-11506359 on Build GRCh38
rs763406367 Chr.19: 11506276-11506276 on Build GRCh38
rs780615143 Chr.19: 11506400-11506400 on Build GRCh38
rs745526995 Chr.19: 11506317-11506317 on Build GRCh38
rs779487480 Chr.19: 11506284-11506284 on Build GRCh38
rs775487934 Chr.19: 11506358-11506358 on Build GRCh38
rs763975201 Chr.19: 11506369-11506369 on Build GRCh38
rs138308187 Chr.19: 11506244-11506244 on Build GRCh38
rs150149179 Chr.19: 11506234-11506234 on Build GRCh38
rs778663665 Chr.19: 11506237-11506237 on Build GRCh38
rs779857790 Chr.19: 11506394-11506394 on Build GRCh38
rs775716908 Chr.19: 11506427-11506427 on Build GRCh38
rs763177733 Chr.19: 11506338-11506338 on Build GRCh38
rs35513404 Chr.19: 11506258-11506258 on Build GRCh38
rs779687690 Chr.19: 11506323-11506323 on Build GRCh38
rs749160236 Chr.19: 11506230-11506230 on Build GRCh38
rs754557544 Chr.19: 11506398-11506398 on Build GRCh38

View Full Product Details