Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37846208-37846465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLT3
Entrez Gene ID
6990
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
RP3, TCTE1L, TCTEX1L
UniGene
Hs.446392
Gene Chromosome Location
Chr.X: 37838836-37847636 on Build GRCh38
Transcripts
NM_006520.2

SNP Detail

SNP ID Chromosome Location
rs35612238 Chr.X: 37846359-37846359 on Build GRCh38
rs374042272 Chr.X: 37846306-37846306 on Build GRCh38
rs139261541 Chr.X: 37846391-37846391 on Build GRCh38
rs930974194 Chr.X: 37846426-37846426 on Build GRCh38
rs753095836 Chr.X: 37846364-37846364 on Build GRCh38
rs565732935 Chr.X: 37846416-37846416 on Build GRCh38
rs761103838 Chr.X: 37846412-37846412 on Build GRCh38
rs757569933 Chr.X: 37846326-37846326 on Build GRCh38
rs867332631 Chr.X: 37846355-37846355 on Build GRCh38
rs972488613 Chr.X: 37846379-37846379 on Build GRCh38
rs867384404 Chr.X: 37846368-37846368 on Build GRCh38
rs370848392 Chr.X: 37846305-37846305 on Build GRCh38
rs779916659 Chr.X: 37846363-37846363 on Build GRCh38
rs201856049 Chr.X: 37846289-37846289 on Build GRCh38
rs757952914 Chr.X: 37846314-37846314 on Build GRCh38

View Full Product Details