Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134773643-134773904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX46
Entrez Gene ID
9879
Gene Name
DEAD-box helicase 46
Gene Aliases
PRPF5, Prp5
UniGene
Hs.406549
Gene Chromosome Location
Chr.5: 134758734-134831138 on Build GRCh38
Transcripts
NM_001300860.1 NM_014829.3 NR_125341.1 XM_011543758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3850205 c.607G>A p.D203N Chr.5: 134773855-134773855 on Build GRCh38
5564702 c.511C>T p.R171* Chr.5: 134773759-134773759 on Build GRCh38
1060932 c.568G>A p.E190K Chr.5: 134773816-134773816 on Build GRCh38
1060931 c.448-1G>T p.? Chr.5: 134773695-134773695 on Build GRCh38
4155454 c.573G>C p.E191D Chr.5: 134773821-134773821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768077610 Chr.5: 134773692-134773692 on Build GRCh38
rs752957140 Chr.5: 134773771-134773771 on Build GRCh38
rs763637539 Chr.5: 134773794-134773796 on Build GRCh38
rs755606313 Chr.5: 134773734-134773734 on Build GRCh38
rs1029975224 Chr.5: 134773782-134773782 on Build GRCh38
rs374470384 Chr.5: 134773764-134773764 on Build GRCh38
rs140064519 Chr.5: 134773815-134773815 on Build GRCh38
rs778014053 Chr.5: 134773806-134773806 on Build GRCh38
rs763758010 Chr.5: 134773689-134773689 on Build GRCh38
rs770003459 Chr.5: 134773866-134773867 on Build GRCh38
rs759439661 Chr.5: 134773854-134773854 on Build GRCh38
rs368446004 Chr.5: 134773864-134773864 on Build GRCh38
rs756723024 Chr.5: 134773701-134773701 on Build GRCh38
rs997156515 Chr.5: 134773714-134773714 on Build GRCh38
rs757969298 Chr.5: 134773681-134773681 on Build GRCh38
rs113321050 Chr.5: 134773685-134773685 on Build GRCh38
rs776758841 Chr.5: 134773823-134773823 on Build GRCh38
rs747210353 Chr.5: 134773814-134773814 on Build GRCh38
rs772343005 Chr.5: 134773800-134773800 on Build GRCh38
rs748594950 Chr.5: 134773779-134773779 on Build GRCh38
rs759674426 Chr.5: 134773671-134773674 on Build GRCh38
rs754381622 Chr.5: 134773725-134773725 on Build GRCh38
rs756456760 Chr.5: 134773792-134773792 on Build GRCh38
rs1008856456 Chr.5: 134773698-134773698 on Build GRCh38
rs376220576 Chr.5: 134773707-134773707 on Build GRCh38
rs35026423 Chr.5: 134773804-134773804 on Build GRCh38

View Full Product Details