Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41983466-41983735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC7
Entrez Gene ID
7266
Gene Name
DnaJ heat shock protein family (Hsp40) member C7
Gene Aliases
DJ11, DJC7, TPR2, TTC2
UniGene
Hs.500156
Gene Chromosome Location
Chr.17: 41976421-42021371 on Build GRCh38
Transcripts
NM_001144766.2 NM_003315.3 NR_029431.1 XM_011525167.2 XM_011525168.2 XM_011525169.2 XM_017024994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3188227 c.1036G>A p.E346K Chr.17: 41983611-41983611 on Build GRCh38
3188221 c.1073A>T p.E358V Chr.17: 41983574-41983574 on Build GRCh38
706331 c.981-2A>G p.? Chr.17: 41983638-41983638 on Build GRCh38
6146452 c.1025A>T p.E342V Chr.17: 41983592-41983592 on Build GRCh38
3396269 c.1084+5G>T p.? Chr.17: 41983558-41983558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782178627 Chr.17: 41983594-41983594 on Build GRCh38
rs374510378 Chr.17: 41983527-41983527 on Build GRCh38
rs782265836 Chr.17: 41983682-41983682 on Build GRCh38
rs937642526 Chr.17: 41983703-41983703 on Build GRCh38
rs550163769 Chr.17: 41983509-41983509 on Build GRCh38
rs141872445 Chr.17: 41983651-41983651 on Build GRCh38
rs782552563 Chr.17: 41983661-41983661 on Build GRCh38
rs973441757 Chr.17: 41983548-41983548 on Build GRCh38
rs138726127 Chr.17: 41983555-41983555 on Build GRCh38
rs782128553 Chr.17: 41983631-41983631 on Build GRCh38
rs1008977535 Chr.17: 41983513-41983513 on Build GRCh38
rs782377882 Chr.17: 41983686-41983686 on Build GRCh38
rs116160290 Chr.17: 41983662-41983662 on Build GRCh38
rs367848406 Chr.17: 41983601-41983601 on Build GRCh38
rs781958228 Chr.17: 41983584-41983584 on Build GRCh38
rs374329124 Chr.17: 41983614-41983614 on Build GRCh38
rs781789611 Chr.17: 41983654-41983654 on Build GRCh38
rs782296297 Chr.17: 41983529-41983529 on Build GRCh38
rs184359355 Chr.17: 41983644-41983644 on Build GRCh38
rs782208820 Chr.17: 41983518-41983518 on Build GRCh38
rs370163280 Chr.17: 41983560-41983560 on Build GRCh38
rs782465285 Chr.17: 41983550-41983550 on Build GRCh38
rs781840020 Chr.17: 41983543-41983543 on Build GRCh38
rs782569134 Chr.17: 41983519-41983519 on Build GRCh38
rs375324751 Chr.17: 41983582-41983582 on Build GRCh38
rs200375131 Chr.17: 41983546-41983546 on Build GRCh38
rs782758339 Chr.17: 41983633-41983633 on Build GRCh38
rs781836096 Chr.17: 41983642-41983642 on Build GRCh38

View Full Product Details