Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6577724-6577977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD4
Entrez Gene ID
1108
Gene Name
chromodomain helicase DNA binding protein 4
Gene Aliases
CHD-4, Mi-2b, Mi2-BETA
UniGene
Hs.162233
Gene Chromosome Location
Chr.12: 6570082-6607439 on Build GRCh38
Transcripts
NM_001273.3 NM_001297553.1 XM_005253668.3 XM_006718958.1 XM_006718959.1 XM_006718960.1 XM_006718961.2 XM_006718962.1 XM_017018725.1 XM_017018726.1 XM_017018727.1 XM_017018728.1 XM_017018729.1 XM_017018730.1 XM_017018731.1 XM_017018732.1 XM_017018733.1 XM_017018734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4749355 c.5234G>T p.G1745V Chr.12: 6577912-6577912 on Build GRCh38
6627924 c.5401A>G p.N1801D Chr.12: 6577829-6577829 on Build GRCh38
6627918 c.5399G>A p.G1800D Chr.12: 6577831-6577831 on Build GRCh38
4866909 c.5360A>G p.K1787R Chr.12: 6577786-6577786 on Build GRCh38
1248213 c.5423A>C p.K1808T Chr.12: 6577807-6577807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747768498 Chr.12: 6577836-6577836 on Build GRCh38
rs776068155 Chr.12: 6577778-6577779 on Build GRCh38
rs367790494 Chr.12: 6577942-6577942 on Build GRCh38
rs781178939 Chr.12: 6577835-6577835 on Build GRCh38
rs750402336 Chr.12: 6577877-6577877 on Build GRCh38
rs768785035 Chr.12: 6577776-6577776 on Build GRCh38
rs755002159 Chr.12: 6577904-6577904 on Build GRCh38
rs201011723 Chr.12: 6577920-6577920 on Build GRCh38
rs761984548 Chr.12: 6577876-6577876 on Build GRCh38
rs71584868 Chr.12: 6577854-6577854 on Build GRCh38
rs868070133 Chr.12: 6577796-6577796 on Build GRCh38
rs766427424 Chr.12: 6577890-6577890 on Build GRCh38
rs71584867 Chr.12: 6577857-6577857 on Build GRCh38
rs74470219 Chr.12: 6577758-6577758 on Build GRCh38
rs377173786 Chr.12: 6577840-6577840 on Build GRCh38
rs371006626 Chr.12: 6577777-6577777 on Build GRCh38
rs763189593 Chr.12: 6577945-6577945 on Build GRCh38
rs932499522 Chr.12: 6577768-6577771 on Build GRCh38
rs758499850 Chr.12: 6577880-6577880 on Build GRCh38
rs747010374 Chr.12: 6577812-6577812 on Build GRCh38
rs764421286 Chr.12: 6577866-6577866 on Build GRCh38
rs370626030 Chr.12: 6577914-6577914 on Build GRCh38
rs761229855 Chr.12: 6577865-6577865 on Build GRCh38
rs747314906 Chr.12: 6577938-6577938 on Build GRCh38
rs529117126 Chr.12: 6577784-6577784 on Build GRCh38
rs777021265 Chr.12: 6577873-6577873 on Build GRCh38
rs755667676 Chr.12: 6577932-6577932 on Build GRCh38
rs769476360 Chr.12: 6577861-6577861 on Build GRCh38
rs377002874 Chr.12: 6577762-6577762 on Build GRCh38
rs758210504 Chr.12: 6577778-6577778 on Build GRCh38
rs751516106 Chr.12: 6577896-6577896 on Build GRCh38
rs772550482 Chr.12: 6577842-6577842 on Build GRCh38
rs368491094 Chr.12: 6577779-6577779 on Build GRCh38
rs371408838 Chr.12: 6577821-6577821 on Build GRCh38

View Full Product Details