Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103426946-103427219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF13
Entrez Gene ID
25879
Gene Name
DDB1 and CUL4 associated factor 13
Gene Aliases
GM83, HSPC064, WDSOF1
UniGene
Hs.532265
Gene Chromosome Location
Chr.8: 103414714-103443453 on Build GRCh38
Transcripts
NM_015420.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3269816 c.961G>T p.A321S Chr.8: 103427133-103427133 on Build GRCh38
6247251 c.970G>T p.A324S Chr.8: 103427142-103427142 on Build GRCh38
4587706 c.961G>A p.A321T Chr.8: 103427133-103427133 on Build GRCh38
4950410 c.1040G>T p.G347V Chr.8: 103427212-103427212 on Build GRCh38
4186091 c.1045G>T p.D349Y Chr.8: 103427217-103427217 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754945687 Chr.8: 103427077-103427077 on Build GRCh38
rs763042484 Chr.8: 103427064-103427064 on Build GRCh38
rs907395005 Chr.8: 103427068-103427068 on Build GRCh38
rs1035498260 Chr.8: 103426994-103426994 on Build GRCh38
rs992216365 Chr.8: 103427015-103427015 on Build GRCh38
rs769347024 Chr.8: 103427158-103427158 on Build GRCh38
rs568236562 Chr.8: 103427160-103427160 on Build GRCh38
rs753675110 Chr.8: 103427027-103427027 on Build GRCh38
rs748540892 Chr.8: 103427161-103427161 on Build GRCh38
rs773396420 Chr.8: 103427164-103427164 on Build GRCh38
rs974084252 Chr.8: 103427046-103427046 on Build GRCh38
rs778609059 Chr.8: 103427154-103427154 on Build GRCh38
rs774479851 Chr.8: 103427186-103427186 on Build GRCh38
rs983453478 Chr.8: 103427099-103427099 on Build GRCh38
rs1027530882 Chr.8: 103427156-103427156 on Build GRCh38
rs751613260 Chr.8: 103427076-103427076 on Build GRCh38
rs777363133 Chr.8: 103427106-103427106 on Build GRCh38
rs187260078 Chr.8: 103427058-103427058 on Build GRCh38
rs969352153 Chr.8: 103427045-103427045 on Build GRCh38
rs960161656 Chr.8: 103427011-103427011 on Build GRCh38
rs755950053 Chr.8: 103427104-103427104 on Build GRCh38
rs748993515 Chr.8: 103427122-103427122 on Build GRCh38
rs550078607 Chr.8: 103427080-103427080 on Build GRCh38
rs766465691 Chr.8: 103427073-103427073 on Build GRCh38
rs145609503 Chr.8: 103427157-103427157 on Build GRCh38
rs763209111 Chr.8: 103427176-103427176 on Build GRCh38
rs535190221 Chr.8: 103427147-103427147 on Build GRCh38
rs148884091 Chr.8: 103427163-103427163 on Build GRCh38
rs756947557 Chr.8: 103427141-103427141 on Build GRCh38
rs374751047 Chr.8: 103427087-103427087 on Build GRCh38

View Full Product Details