Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1773662-1773907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME2
Entrez Gene ID
197342
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
SLX2B, gs125
UniGene
Hs.7247
Gene Chromosome Location
Chr.16: 1773222-1776714 on Build GRCh38
Transcripts
NM_001257370.1 XM_017023025.1 XM_017023026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6299653 c.314G>A p.R105H Chr.16: 1773771-1773771 on Build GRCh38
6397129 c.344T>A p.L115Q Chr.16: 1773801-1773801 on Build GRCh38
7366182 c.304T>C p.C102R Chr.16: 1773761-1773761 on Build GRCh38
4852963 c.319G>C p.E107Q Chr.16: 1773776-1773776 on Build GRCh38
5740515 c.286G>A p.A96T Chr.16: 1773743-1773743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755764387 Chr.16: 1773694-1773694 on Build GRCh38
rs767363303 Chr.16: 1773763-1773763 on Build GRCh38
rs763128723 Chr.16: 1773734-1773734 on Build GRCh38
rs201737681 Chr.16: 1773739-1773739 on Build GRCh38
rs776920899 Chr.16: 1773720-1773720 on Build GRCh38
rs774425457 Chr.16: 1773873-1773873 on Build GRCh38
rs1028445254 Chr.16: 1773823-1773823 on Build GRCh38
rs778471994 Chr.16: 1773810-1773810 on Build GRCh38
rs750205139 Chr.16: 1773777-1773777 on Build GRCh38
rs747546404 Chr.16: 1773715-1773715 on Build GRCh38
rs920676787 Chr.16: 1773752-1773752 on Build GRCh38
rs558383127 Chr.16: 1773682-1773682 on Build GRCh38
rs547679861 Chr.16: 1773839-1773839 on Build GRCh38
rs76650990 Chr.16: 1773692-1773692 on Build GRCh38
rs886910511 Chr.16: 1773789-1773789 on Build GRCh38
rs751652697 Chr.16: 1773746-1773746 on Build GRCh38
rs779884399 Chr.16: 1773698-1773698 on Build GRCh38
rs199761703 Chr.16: 1773727-1773727 on Build GRCh38
rs759897366 Chr.16: 1773722-1773722 on Build GRCh38
rs996937332 Chr.16: 1773688-1773688 on Build GRCh38
rs917501982 Chr.16: 1773881-1773881 on Build GRCh38
rs1040041297 Chr.16: 1773771-1773771 on Build GRCh38
rs1031403402 Chr.16: 1773853-1773853 on Build GRCh38
rs761755726 Chr.16: 1773762-1773762 on Build GRCh38
rs780043716 Chr.16: 1773786-1773786 on Build GRCh38
rs770118349 Chr.16: 1773836-1773836 on Build GRCh38
rs573283501 Chr.16: 1773827-1773827 on Build GRCh38
rs567315180 Chr.16: 1773864-1773864 on Build GRCh38
rs1040947078 Chr.16: 1773800-1773800 on Build GRCh38
rs773774708 Chr.16: 1773714-1773714 on Build GRCh38
rs945752344 Chr.16: 1773758-1773758 on Build GRCh38
rs761833968 Chr.16: 1773874-1773874 on Build GRCh38
rs901114865 Chr.16: 1773816-1773816 on Build GRCh38
rs557282646 Chr.16: 1773737-1773738 on Build GRCh38
rs768190872 Chr.16: 1773708-1773708 on Build GRCh38
rs961733474 Chr.16: 1773684-1773684 on Build GRCh38
rs755542062 Chr.16: 1773818-1773818 on Build GRCh38
rs948735777 Chr.16: 1773733-1773733 on Build GRCh38
rs763227586 Chr.16: 1773845-1773845 on Build GRCh38
rs746179767 Chr.16: 1773832-1773832 on Build GRCh38
rs924742128 Chr.16: 1773711-1773711 on Build GRCh38
rs781684913 Chr.16: 1773828-1773828 on Build GRCh38
rs771398630 Chr.16: 1773716-1773716 on Build GRCh38
rs765401210 Chr.16: 1773723-1773723 on Build GRCh38
rs755997861 Chr.16: 1773785-1773785 on Build GRCh38
rs756465784 Chr.16: 1773738-1773746 on Build GRCh38
rs996083815 Chr.16: 1773819-1773819 on Build GRCh38
rs993072488 Chr.16: 1773721-1773721 on Build GRCh38
rs374315507 Chr.16: 1773766-1773766 on Build GRCh38
rs754653019 Chr.16: 1773794-1773794 on Build GRCh38
rs956189417 Chr.16: 1773855-1773855 on Build GRCh38
rs917201339 Chr.16: 1773693-1773693 on Build GRCh38
rs531318985 Chr.16: 1773702-1773702 on Build GRCh38
rs753510592 Chr.16: 1773790-1773790 on Build GRCh38

View Full Product Details