Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 99015388-99015623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL15A1
Entrez Gene ID
1306
Gene Name
collagen type XV alpha 1 chain
Gene Aliases
-
UniGene
Hs.409034
Gene Chromosome Location
Chr.9: 98943392-99070792 on Build GRCh38
Transcripts
NM_001855.4 XM_011518214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7176864 c.1375T>G p.L459V Chr.9: 99015438-99015438 on Build GRCh38
5699463 c.1502C>T p.S501F Chr.9: 99015565-99015565 on Build GRCh38
6373616 c.1460C>A p.A487D Chr.9: 99015523-99015523 on Build GRCh38
3367471 c.1449A>C p.T483T Chr.9: 99015512-99015512 on Build GRCh38
3726847 c.1481_1482insA p.E496fs*1 Chr.9: 99015544-99015545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202155886 Chr.9: 99015552-99015552 on Build GRCh38
rs763285450 Chr.9: 99015469-99015469 on Build GRCh38
rs368294690 Chr.9: 99015475-99015475 on Build GRCh38
rs760232458 Chr.9: 99015599-99015599 on Build GRCh38
rs145278600 Chr.9: 99015553-99015553 on Build GRCh38
rs149002114 Chr.9: 99015525-99015525 on Build GRCh38
rs780113490 Chr.9: 99015564-99015564 on Build GRCh38
rs145891402 Chr.9: 99015462-99015462 on Build GRCh38
rs1056786986 Chr.9: 99015575-99015575 on Build GRCh38
rs768653244 Chr.9: 99015452-99015452 on Build GRCh38
rs759796106 Chr.9: 99015585-99015585 on Build GRCh38
rs774726276 Chr.9: 99015495-99015495 on Build GRCh38
rs760047166 Chr.9: 99015497-99015497 on Build GRCh38
rs773565512 Chr.9: 99015466-99015466 on Build GRCh38
rs753511937 Chr.9: 99015535-99015535 on Build GRCh38
rs371422739 Chr.9: 99015601-99015601 on Build GRCh38
rs776806733 Chr.9: 99015458-99015458 on Build GRCh38
rs771232853 Chr.9: 99015578-99015578 on Build GRCh38
rs757013439 Chr.9: 99015540-99015540 on Build GRCh38
rs374041558 Chr.9: 99015576-99015576 on Build GRCh38
rs771272092 Chr.9: 99015476-99015476 on Build GRCh38
rs973831064 Chr.9: 99015547-99015547 on Build GRCh38
rs200414003 Chr.9: 99015532-99015532 on Build GRCh38
rs778155382 Chr.9: 99015573-99015573 on Build GRCh38
rs772607127 Chr.9: 99015588-99015588 on Build GRCh38
rs374363919 Chr.9: 99015445-99015445 on Build GRCh38
rs956050078 Chr.9: 99015425-99015425 on Build GRCh38
rs150243247 Chr.9: 99015433-99015433 on Build GRCh38
rs1029921600 Chr.9: 99015451-99015451 on Build GRCh38
rs778700512 Chr.9: 99015549-99015549 on Build GRCh38
rs200379044 Chr.9: 99015461-99015461 on Build GRCh38
rs774808163 Chr.9: 99015412-99015412 on Build GRCh38
rs113429679 Chr.9: 99015508-99015508 on Build GRCh38
rs767034635 Chr.9: 99015499-99015499 on Build GRCh38
rs760146673 Chr.9: 99015530-99015530 on Build GRCh38
rs748308539 Chr.9: 99015567-99015567 on Build GRCh38
rs759515802 Chr.9: 99015599-99015600 on Build GRCh38
rs1000253753 Chr.9: 99015410-99015410 on Build GRCh38
rs745757992 Chr.9: 99015450-99015450 on Build GRCh38
rs936995411 Chr.9: 99015567-99015570 on Build GRCh38
rs1033458521 Chr.9: 99015424-99015424 on Build GRCh38
rs777380955 Chr.9: 99015435-99015435 on Build GRCh38
rs763799487 Chr.9: 99015534-99015534 on Build GRCh38
rs757066128 Chr.9: 99015448-99015448 on Build GRCh38
rs769982025 Chr.9: 99015571-99015571 on Build GRCh38
rs911360407 Chr.9: 99015471-99015471 on Build GRCh38
rs752376118 Chr.9: 99015426-99015426 on Build GRCh38
rs150779155 Chr.9: 99015584-99015584 on Build GRCh38
rs550619128 Chr.9: 99015449-99015449 on Build GRCh38
rs1036965253 Chr.9: 99015596-99015596 on Build GRCh38

View Full Product Details