Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50479660-50479925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDC
Entrez Gene ID
1644
Gene Name
dopa decarboxylase
Gene Aliases
AADC
UniGene
Hs.359698
Gene Chromosome Location
Chr.7: 50458436-50565460 on Build GRCh38
Transcripts
NM_000790.3 NM_001082971.1 NM_001242886.1 NM_001242887.1 NM_001242888.1 NM_001242889.1 XM_005271745.4 XM_011515161.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
223169 c.1015G>A p.D339N Chr.7: 50479793-50479793 on Build GRCh38
5042356 c.970G>A p.G324R Chr.7: 50479838-50479838 on Build GRCh38
5603351 c.991A>T p.T331S Chr.7: 50479817-50479817 on Build GRCh38
1451341 c.956G>T p.R319I Chr.7: 50479852-50479852 on Build GRCh38
3265500 c.968C>T p.T323M Chr.7: 50479840-50479840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779203355 Chr.7: 50479900-50479900 on Build GRCh38
rs11575457 Chr.7: 50479886-50479886 on Build GRCh38
rs946105201 Chr.7: 50479781-50479781 on Build GRCh38
rs750269360 Chr.7: 50479778-50479778 on Build GRCh38
rs755084437 Chr.7: 50479800-50479800 on Build GRCh38
rs768264090 Chr.7: 50479835-50479835 on Build GRCh38
rs758505664 Chr.7: 50479791-50479791 on Build GRCh38
rs760243492 Chr.7: 50479746-50479746 on Build GRCh38
rs140562157 Chr.7: 50479840-50479840 on Build GRCh38
rs538476047 Chr.7: 50479883-50479883 on Build GRCh38
rs766157857 Chr.7: 50479871-50479871 on Build GRCh38
rs373966178 Chr.7: 50479798-50479798 on Build GRCh38
rs759081023 Chr.7: 50479742-50479742 on Build GRCh38
rs764055985 Chr.7: 50479751-50479751 on Build GRCh38
rs942973348 Chr.7: 50479879-50479879 on Build GRCh38
rs771564251 Chr.7: 50479821-50479821 on Build GRCh38
rs759803257 Chr.7: 50479757-50479757 on Build GRCh38
rs536353640 Chr.7: 50479704-50479704 on Build GRCh38
rs368472358 Chr.7: 50479885-50479885 on Build GRCh38
rs751797399 Chr.7: 50479876-50479876 on Build GRCh38
rs576576188 Chr.7: 50479737-50479737 on Build GRCh38
rs75490403 Chr.7: 50479684-50479684 on Build GRCh38
rs886203920 Chr.7: 50479904-50479904 on Build GRCh38
rs751531676 Chr.7: 50479799-50479799 on Build GRCh38
rs1004611651 Chr.7: 50479905-50479905 on Build GRCh38
rs958995120 Chr.7: 50479711-50479711 on Build GRCh38
rs756656475 Chr.7: 50479814-50479814 on Build GRCh38
rs755244033 Chr.7: 50479878-50479878 on Build GRCh38
rs775212133 Chr.7: 50479822-50479822 on Build GRCh38
rs772828164 Chr.7: 50479859-50479859 on Build GRCh38
rs766640988 Chr.7: 50479794-50479794 on Build GRCh38
rs776704659 Chr.7: 50479764-50479764 on Build GRCh38
rs757636797 Chr.7: 50479889-50479889 on Build GRCh38
rs748632382 Chr.7: 50479803-50479803 on Build GRCh38
rs561268905 Chr.7: 50479718-50479718 on Build GRCh38
rs749547440 Chr.7: 50479887-50479887 on Build GRCh38
rs761895984 Chr.7: 50479775-50479775 on Build GRCh38
rs781308256 Chr.7: 50479801-50479801 on Build GRCh38
rs940802845 Chr.7: 50479831-50479831 on Build GRCh38
rs971473432 Chr.7: 50479692-50479692 on Build GRCh38
rs778072349 Chr.7: 50479815-50479815 on Build GRCh38
rs765294373 Chr.7: 50479852-50479852 on Build GRCh38
rs746705439 Chr.7: 50479827-50479827 on Build GRCh38
rs188088947 Chr.7: 50479861-50479861 on Build GRCh38
rs761808166 Chr.7: 50479849-50479849 on Build GRCh38
rs774812091 Chr.7: 50479745-50479745 on Build GRCh38
rs765098329 Chr.7: 50479777-50479777 on Build GRCh38
rs749684919 Chr.7: 50479819-50479819 on Build GRCh38
rs981957907 Chr.7: 50479700-50479700 on Build GRCh38

View Full Product Details