Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12477611-12477882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP16
Entrez Gene ID
80824
Gene Name
dual specificity phosphatase 16
Gene Aliases
MKP-7, MKP7
UniGene
Hs.536535
Gene Chromosome Location
Chr.12: 12473282-12562839 on Build GRCh38
Transcripts
NM_030640.2 XM_005253488.2 XM_006719155.2 XM_011520856.1 XM_011520857.1 XM_017019988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415963 c.1040C>T p.S347L Chr.12: 12477791-12477791 on Build GRCh38
4998122 c.1159G>T p.A387S Chr.12: 12477672-12477672 on Build GRCh38
5027542 c.1204C>G p.L402V Chr.12: 12477627-12477627 on Build GRCh38
5428348 c.1080G>A p.V360V Chr.12: 12477751-12477751 on Build GRCh38
2227591 c.1189A>G p.K397E Chr.12: 12477642-12477642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773448870 Chr.12: 12477851-12477851 on Build GRCh38
rs374755640 Chr.12: 12477725-12477725 on Build GRCh38
rs1050694180 Chr.12: 12477829-12477829 on Build GRCh38
rs761079204 Chr.12: 12477763-12477763 on Build GRCh38
rs777460915 Chr.12: 12477672-12477672 on Build GRCh38
rs200541112 Chr.12: 12477739-12477740 on Build GRCh38
rs758026767 Chr.12: 12477655-12477655 on Build GRCh38
rs762082661 Chr.12: 12477756-12477756 on Build GRCh38
rs757949409 Chr.12: 12477728-12477728 on Build GRCh38
rs756271288 Chr.12: 12477748-12477753 on Build GRCh38
rs781189422 Chr.12: 12477738-12477738 on Build GRCh38
rs370409500 Chr.12: 12477768-12477768 on Build GRCh38
rs756910551 Chr.12: 12477736-12477736 on Build GRCh38
rs576308597 Chr.12: 12477754-12477754 on Build GRCh38
rs773610264 Chr.12: 12477760-12477760 on Build GRCh38
rs755958341 Chr.12: 12477805-12477805 on Build GRCh38
rs754759731 Chr.12: 12477744-12477746 on Build GRCh38
rs59897494 Chr.12: 12477758-12477759 on Build GRCh38
rs3809199 Chr.12: 12477735-12477735 on Build GRCh38
rs776806192 Chr.12: 12477860-12477860 on Build GRCh38
rs756649680 Chr.12: 12477717-12477717 on Build GRCh38
rs771045052 Chr.12: 12477794-12477796 on Build GRCh38
rs933842223 Chr.12: 12477682-12477682 on Build GRCh38
rs777848642 Chr.12: 12477826-12477826 on Build GRCh38
rs142947418 Chr.12: 12477732-12477740 on Build GRCh38
rs777380063 Chr.12: 12477731-12477731 on Build GRCh38
rs370672836 Chr.12: 12477712-12477712 on Build GRCh38
rs138331584 Chr.12: 12477809-12477809 on Build GRCh38
rs759896794 Chr.12: 12477772-12477772 on Build GRCh38
rs748196842 Chr.12: 12477832-12477832 on Build GRCh38
rs144222400 Chr.12: 12477802-12477802 on Build GRCh38
rs766259105 Chr.12: 12477739-12477744 on Build GRCh38
rs200271649 Chr.12: 12477748-12477751 on Build GRCh38
rs142652742 Chr.12: 12477706-12477706 on Build GRCh38
rs201941751 Chr.12: 12477742-12477746 on Build GRCh38
rs772280915 Chr.12: 12477759-12477759 on Build GRCh38
rs570722524 Chr.12: 12477681-12477681 on Build GRCh38
rs200573264 Chr.12: 12477707-12477707 on Build GRCh38
rs761414777 Chr.12: 12477731-12477732 on Build GRCh38
rs529933489 Chr.12: 12477650-12477650 on Build GRCh38
rs762478161 Chr.12: 12477735-12477741 on Build GRCh38
rs779032871 Chr.12: 12477831-12477831 on Build GRCh38
rs768026556 Chr.12: 12477731-12477739 on Build GRCh38
rs538158127 Chr.12: 12477684-12477684 on Build GRCh38
rs975340665 Chr.12: 12477664-12477664 on Build GRCh38
rs760620388 Chr.12: 12477742-12477751 on Build GRCh38
rs112390593 Chr.12: 12477673-12477673 on Build GRCh38
rs767817494 Chr.12: 12477638-12477638 on Build GRCh38
rs781313335 Chr.12: 12477744-12477750 on Build GRCh38
rs749592040 Chr.12: 12477753-12477754 on Build GRCh38
rs763708055 Chr.12: 12477786-12477786 on Build GRCh38
rs372273974 Chr.12: 12477827-12477827 on Build GRCh38
rs777687323 Chr.12: 12477750-12477752 on Build GRCh38
rs771333805 Chr.12: 12477858-12477858 on Build GRCh38
rs767885144 Chr.12: 12477709-12477709 on Build GRCh38
rs760095663 Chr.12: 12477728-12477729 on Build GRCh38
rs775472122 Chr.12: 12477745-12477745 on Build GRCh38
rs955439629 Chr.12: 12477803-12477803 on Build GRCh38
rs61757366 Chr.12: 12477744-12477744 on Build GRCh38
rs752199007 Chr.12: 12477727-12477727 on Build GRCh38
rs367623951 Chr.12: 12477804-12477804 on Build GRCh38
rs557753282 Chr.12: 12477753-12477753 on Build GRCh38
rs2111269 Chr.12: 12477751-12477751 on Build GRCh38
rs760772589 Chr.12: 12477853-12477853 on Build GRCh38
rs775499226 Chr.12: 12477695-12477695 on Build GRCh38
rs756589989 Chr.12: 12477646-12477646 on Build GRCh38
rs141269948 Chr.12: 12477855-12477855 on Build GRCh38
rs750138441 Chr.12: 12477735-12477737 on Build GRCh38
rs114832912 Chr.12: 12477769-12477769 on Build GRCh38
rs3055727 Chr.12: 12477742-12477742 on Build GRCh38
rs150065682 Chr.12: 12477694-12477694 on Build GRCh38
rs751098837 Chr.12: 12477801-12477801 on Build GRCh38
rs574643405 Chr.12: 12477740-12477741 on Build GRCh38
rs763053694 Chr.12: 12477703-12477703 on Build GRCh38
rs772329969 Chr.12: 12477846-12477846 on Build GRCh38
rs527241198 Chr.12: 12477639-12477639 on Build GRCh38
rs140147069 Chr.12: 12477691-12477691 on Build GRCh38
rs747693297 Chr.12: 12477651-12477651 on Build GRCh38

View Full Product Details