Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120400697-120400956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP2
Entrez Gene ID
55755
Gene Name
CDK5 regulatory subunit associated protein 2
Gene Aliases
C48, Cep215, MCPH3
UniGene
Hs.269560
Gene Chromosome Location
Chr.9: 120388869-120580170 on Build GRCh38
Transcripts
NM_001011649.2 NM_001272039.1 NM_018249.5 NR_073554.1 NR_073555.1 NR_073556.1 NR_073557.1 NR_073558.1 XM_006717182.1 XM_006717185.1 XM_011518860.1 XM_017014921.1 XM_017014922.1 XM_017014923.1 XM_017014924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3926082 c.5377G>A p.A1793T Chr.9: 120400816-120400816 on Build GRCh38
3982721 c.5418C>A p.P1806P Chr.9: 120400775-120400775 on Build GRCh38
5721396 c.5416C>T p.P1806S Chr.9: 120400777-120400777 on Build GRCh38
4152017 c.5352C>T p.S1784S Chr.9: 120400841-120400841 on Build GRCh38
4499479 c.5436C>T p.P1812P Chr.9: 120400757-120400757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs587783395 Chr.9: 120400834-120400834 on Build GRCh38
rs769913178 Chr.9: 120400790-120400790 on Build GRCh38
rs750087275 Chr.9: 120400913-120400913 on Build GRCh38
rs919150866 Chr.9: 120400781-120400781 on Build GRCh38
rs991061168 Chr.9: 120400829-120400829 on Build GRCh38
rs374808413 Chr.9: 120400918-120400918 on Build GRCh38
rs752673678 Chr.9: 120400827-120400827 on Build GRCh38
rs145402135 Chr.9: 120400838-120400838 on Build GRCh38
rs148857536 Chr.9: 120400875-120400875 on Build GRCh38
rs774322006 Chr.9: 120400820-120400820 on Build GRCh38
rs753295890 Chr.9: 120400833-120400833 on Build GRCh38
rs774698258 Chr.9: 120400892-120400892 on Build GRCh38
rs767985869 Chr.9: 120400718-120400718 on Build GRCh38
rs773359220 Chr.9: 120400802-120400802 on Build GRCh38
rs144163551 Chr.9: 120400823-120400823 on Build GRCh38
rs3739822 Chr.9: 120400775-120400775 on Build GRCh38
rs545543130 Chr.9: 120400780-120400780 on Build GRCh38
rs191994393 Chr.9: 120400896-120400896 on Build GRCh38
rs923647286 Chr.9: 120400881-120400881 on Build GRCh38
rs775085744 Chr.9: 120400901-120400901 on Build GRCh38
rs753022449 Chr.9: 120400762-120400762 on Build GRCh38
rs1035160144 Chr.9: 120400788-120400788 on Build GRCh38
rs994826632 Chr.9: 120400743-120400743 on Build GRCh38
rs770145694 Chr.9: 120400885-120400885 on Build GRCh38
rs748693383 Chr.9: 120400877-120400877 on Build GRCh38
rs778024826 Chr.9: 120400890-120400890 on Build GRCh38
rs751971032 Chr.9: 120400839-120400839 on Build GRCh38
rs761289200 Chr.9: 120400911-120400911 on Build GRCh38
rs564228451 Chr.9: 120400835-120400835 on Build GRCh38
rs752195245 Chr.9: 120400717-120400717 on Build GRCh38
rs763603318 Chr.9: 120400832-120400832 on Build GRCh38
rs147693474 Chr.9: 120400891-120400891 on Build GRCh38
rs142478904 Chr.9: 120400915-120400915 on Build GRCh38
rs769080804 Chr.9: 120400766-120400766 on Build GRCh38
rs747332092 Chr.9: 120400764-120400764 on Build GRCh38
rs375896806 Chr.9: 120400831-120400831 on Build GRCh38
rs587783396 Chr.9: 120400745-120400745 on Build GRCh38
rs370978182 Chr.9: 120400912-120400912 on Build GRCh38
rs756844439 Chr.9: 120400747-120400747 on Build GRCh38
rs781518429 Chr.9: 120400897-120400898 on Build GRCh38
rs759346683 Chr.9: 120400822-120400822 on Build GRCh38
rs771901792 Chr.9: 120400900-120400900 on Build GRCh38
rs771233738 Chr.9: 120400813-120400813 on Build GRCh38
rs760684617 Chr.9: 120400902-120400902 on Build GRCh38
rs763286312 Chr.9: 120400807-120400807 on Build GRCh38
rs771258447 Chr.9: 120400727-120400728 on Build GRCh38
rs753384409 Chr.9: 120400907-120400907 on Build GRCh38
rs780417847 Chr.9: 120400752-120400752 on Build GRCh38
rs755634534 Chr.9: 120400725-120400725 on Build GRCh38
rs372636070 Chr.9: 120400734-120400734 on Build GRCh38
rs763979058 Chr.9: 120400904-120400904 on Build GRCh38

View Full Product Details