Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47736314-47736587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD2
Entrez Gene ID
30846
Gene Name
EH domain containing 2
Gene Aliases
PAST2
UniGene
Hs.744963
Gene Chromosome Location
Chr.19: 47713344-47743134 on Build GRCh38
Transcripts
NM_014601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
380152 c.1048C>T p.P350S Chr.19: 47736501-47736501 on Build GRCh38
6652420 c.1009G>A p.V337I Chr.19: 47736462-47736462 on Build GRCh38
4655782 c.923C>A p.A308D Chr.19: 47736376-47736376 on Build GRCh38
106543 c.1011C>T p.V337V Chr.19: 47736464-47736464 on Build GRCh38
7139446 c.1063G>T p.D355Y Chr.19: 47736516-47736516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs559224950 Chr.19: 47736383-47736383 on Build GRCh38
rs565135733 Chr.19: 47736391-47736391 on Build GRCh38
rs200234195 Chr.19: 47736400-47736400 on Build GRCh38
rs75909126 Chr.19: 47736461-47736461 on Build GRCh38
rs11672953 Chr.19: 47736549-47736549 on Build GRCh38
rs62130744 Chr.19: 47736341-47736341 on Build GRCh38
rs111705699 Chr.19: 47736367-47736367 on Build GRCh38
rs1036353470 Chr.19: 47736352-47736352 on Build GRCh38
rs150829735 Chr.19: 47736347-47736347 on Build GRCh38
rs199706388 Chr.19: 47736389-47736389 on Build GRCh38
rs1003195355 Chr.19: 47736532-47736532 on Build GRCh38
rs536783884 Chr.19: 47736473-47736473 on Build GRCh38
rs201423715 Chr.19: 47736344-47736344 on Build GRCh38
rs764048160 Chr.19: 47736508-47736508 on Build GRCh38
rs760046217 Chr.19: 47736353-47736353 on Build GRCh38
rs768217299 Chr.19: 47736372-47736372 on Build GRCh38
rs369898716 Chr.19: 47736410-47736410 on Build GRCh38
rs776110210 Chr.19: 47736374-47736374 on Build GRCh38
rs199780537 Chr.19: 47736431-47736431 on Build GRCh38
rs766223992 Chr.19: 47736485-47736485 on Build GRCh38
rs751918633 Chr.19: 47736424-47736426 on Build GRCh38
rs200567810 Chr.19: 47736377-47736377 on Build GRCh38
rs561331165 Chr.19: 47736384-47736384 on Build GRCh38
rs754036473 Chr.19: 47736510-47736510 on Build GRCh38
rs372565732 Chr.19: 47736414-47736414 on Build GRCh38
rs747452965 Chr.19: 47736559-47736559 on Build GRCh38
rs755749398 Chr.19: 47736491-47736491 on Build GRCh38
rs138130337 Chr.19: 47736495-47736495 on Build GRCh38
rs758651936 Chr.19: 47736550-47736550 on Build GRCh38
rs1028265447 Chr.19: 47736375-47736375 on Build GRCh38
rs772885356 Chr.19: 47736523-47736523 on Build GRCh38
rs774103640 Chr.19: 47736486-47736486 on Build GRCh38
rs747583344 Chr.19: 47736453-47736453 on Build GRCh38
rs764998477 Chr.19: 47736381-47736381 on Build GRCh38
rs762877549 Chr.19: 47736388-47736388 on Build GRCh38
rs372331248 Chr.19: 47736395-47736395 on Build GRCh38
rs761441836 Chr.19: 47736472-47736472 on Build GRCh38
rs762792858 Chr.19: 47736526-47736526 on Build GRCh38
rs756102458 Chr.19: 47736397-47736397 on Build GRCh38
rs116312342 Chr.19: 47736556-47736556 on Build GRCh38
rs377072399 Chr.19: 47736413-47736413 on Build GRCh38
rs201029505 Chr.19: 47736403-47736403 on Build GRCh38
rs761425944 Chr.19: 47736462-47736462 on Build GRCh38
rs151244683 Chr.19: 47736422-47736422 on Build GRCh38
rs765497591 Chr.19: 47736542-47736542 on Build GRCh38
rs141588825 Chr.19: 47736464-47736464 on Build GRCh38
rs368915920 Chr.19: 47736359-47736359 on Build GRCh38
rs772498278 Chr.19: 47736434-47736434 on Build GRCh38
rs199923224 Chr.19: 47736438-47736438 on Build GRCh38
rs548741584 Chr.19: 47736546-47736546 on Build GRCh38
rs111648404 Chr.19: 47736368-47736368 on Build GRCh38
rs139276620 Chr.19: 47736459-47736459 on Build GRCh38

View Full Product Details