Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 120078338-120078609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L5
Entrez Gene ID
57669
Gene Name
erythrocyte membrane protein band 4.1 like 5
Gene Aliases
BE37, YMO1, YRT
UniGene
Hs.369232
Gene Chromosome Location
Chr.2: 120013028-120179121 on Build GRCh38
Transcripts
NM_001184937.1 NM_001184938.2 NM_001184939.1 NM_020909.3 NR_135920.1 XM_006712651.1 XM_006712652.1 XM_006712653.1 XM_011511529.2 XM_017004567.1 XM_017004568.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4747814 c.715-213_720del219 p.? Chr.2: 120078280-120078498 on Build GRCh38
5911635 c.757G>A p.G253R Chr.2: 120078535-120078535 on Build GRCh38
6654783 c.766_767insT p.E258fs*1 Chr.2: 120078544-120078545 on Build GRCh38
5497647 c.731A>G p.D244G Chr.2: 120078509-120078509 on Build GRCh38
5911636 c.757G>A p.G253R Chr.2: 120078535-120078535 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952835034 Chr.2: 120078559-120078559 on Build GRCh38
rs11677907 Chr.2: 120078382-120078382 on Build GRCh38
rs772917687 Chr.2: 120078520-120078520 on Build GRCh38
rs532480798 Chr.2: 120078431-120078431 on Build GRCh38
rs762802152 Chr.2: 120078523-120078523 on Build GRCh38
rs767728074 Chr.2: 120078558-120078558 on Build GRCh38
rs537905834 Chr.2: 120078437-120078437 on Build GRCh38
rs776078752 Chr.2: 120078504-120078504 on Build GRCh38
rs945261972 Chr.2: 120078471-120078471 on Build GRCh38
rs1028339887 Chr.2: 120078384-120078384 on Build GRCh38
rs751521145 Chr.2: 120078544-120078544 on Build GRCh38
rs376389941 Chr.2: 120078510-120078510 on Build GRCh38
rs749633715 Chr.2: 120078465-120078465 on Build GRCh38
rs199964935 Chr.2: 120078481-120078481 on Build GRCh38
rs768157420 Chr.2: 120078497-120078497 on Build GRCh38
rs867225222 Chr.2: 120078557-120078557 on Build GRCh38
rs752807562 Chr.2: 120078579-120078579 on Build GRCh38
rs920189284 Chr.2: 120078512-120078512 on Build GRCh38
rs775953494 Chr.2: 120078516-120078516 on Build GRCh38
rs937896180 Chr.2: 120078383-120078383 on Build GRCh38
rs961713989 Chr.2: 120078496-120078496 on Build GRCh38
rs761476487 Chr.2: 120078506-120078506 on Build GRCh38
rs1053632402 Chr.2: 120078407-120078407 on Build GRCh38
rs774909828 Chr.2: 120078469-120078469 on Build GRCh38
rs558458059 Chr.2: 120078446-120078446 on Build GRCh38
rs569029005 Chr.2: 120078467-120078467 on Build GRCh38
rs954110618 Chr.2: 120078424-120078424 on Build GRCh38
rs766032836 Chr.2: 120078525-120078525 on Build GRCh38
rs754926603 Chr.2: 120078555-120078555 on Build GRCh38

View Full Product Details