Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96269195-96269468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIAO1
Entrez Gene ID
9391
Gene Name
cytosolic iron-sulfur assembly component 1
Gene Aliases
CIA1, WDR39
UniGene
Hs.12109
Gene Chromosome Location
Chr.2: 96266146-96274179 on Build GRCh38
Transcripts
NM_004804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629159 c.719G>C p.S240T Chr.2: 96269295-96269295 on Build GRCh38
2825374 c.693G>A p.G231G Chr.2: 96269269-96269269 on Build GRCh38
1023860 c.694G>A p.V232M Chr.2: 96269270-96269270 on Build GRCh38
5528698 c.710C>T p.S237F Chr.2: 96269286-96269286 on Build GRCh38
7361320 c.755C>T p.S252L Chr.2: 96269331-96269331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369905973 Chr.2: 96269377-96269377 on Build GRCh38
rs756565539 Chr.2: 96269240-96269240 on Build GRCh38
rs376368442 Chr.2: 96269367-96269367 on Build GRCh38
rs767606370 Chr.2: 96269367-96269368 on Build GRCh38
rs758142894 Chr.2: 96269231-96269231 on Build GRCh38
rs529930540 Chr.2: 96269372-96269372 on Build GRCh38
rs772197431 Chr.2: 96269338-96269338 on Build GRCh38
rs6745581 Chr.2: 96269425-96269425 on Build GRCh38
rs764794024 Chr.2: 96269290-96269290 on Build GRCh38
rs202222774 Chr.2: 96269269-96269269 on Build GRCh38
rs200912319 Chr.2: 96269348-96269348 on Build GRCh38
rs776406200 Chr.2: 96269282-96269282 on Build GRCh38
rs991872675 Chr.2: 96269361-96269361 on Build GRCh38
rs982523554 Chr.2: 96269388-96269388 on Build GRCh38
rs758950152 Chr.2: 96269223-96269223 on Build GRCh38
rs768689225 Chr.2: 96269252-96269252 on Build GRCh38
rs748223775 Chr.2: 96269328-96269328 on Build GRCh38
rs772288953 Chr.2: 96269265-96269265 on Build GRCh38
rs369474100 Chr.2: 96269261-96269261 on Build GRCh38
rs763830889 Chr.2: 96269305-96269305 on Build GRCh38
rs753872609 Chr.2: 96269221-96269221 on Build GRCh38
rs747299720 Chr.2: 96269360-96269360 on Build GRCh38
rs909088566 Chr.2: 96269446-96269446 on Build GRCh38
rs774847204 Chr.2: 96269257-96269258 on Build GRCh38
rs373239060 Chr.2: 96269275-96269275 on Build GRCh38
rs373326907 Chr.2: 96269238-96269238 on Build GRCh38
rs760375256 Chr.2: 96269273-96269273 on Build GRCh38
rs762625261 Chr.2: 96269303-96269303 on Build GRCh38
rs764883846 Chr.2: 96269227-96269227 on Build GRCh38
rs763744880 Chr.2: 96269389-96269389 on Build GRCh38
rs752360148 Chr.2: 96269230-96269230 on Build GRCh38
rs777836690 Chr.2: 96269339-96269339 on Build GRCh38
rs1033085946 Chr.2: 96269281-96269281 on Build GRCh38
rs778938685 Chr.2: 96269299-96269299 on Build GRCh38
rs867841642 Chr.2: 96269267-96269267 on Build GRCh38
rs749850437 Chr.2: 96269242-96269242 on Build GRCh38
rs146638347 Chr.2: 96269286-96269286 on Build GRCh38
rs199619715 Chr.2: 96269279-96269279 on Build GRCh38
rs375053691 Chr.2: 96269241-96269241 on Build GRCh38
rs546763223 Chr.2: 96269433-96269433 on Build GRCh38
rs868312684 Chr.2: 96269268-96269268 on Build GRCh38
rs756411072 Chr.2: 96269307-96269307 on Build GRCh38
rs759492984 Chr.2: 96269373-96269373 on Build GRCh38
rs566996614 Chr.2: 96269434-96269434 on Build GRCh38
rs778899743 Chr.2: 96269326-96269326 on Build GRCh38
rs754327732 Chr.2: 96269237-96269237 on Build GRCh38
rs760031180 Chr.2: 96269367-96269367 on Build GRCh38
rs780507203 Chr.2: 96269320-96269320 on Build GRCh38
rs150250572 Chr.2: 96269291-96269291 on Build GRCh38
rs762535598 Chr.2: 96269387-96269387 on Build GRCh38
rs530725550 Chr.2: 96269258-96269258 on Build GRCh38
rs201052401 Chr.2: 96269321-96269321 on Build GRCh38
rs72937663 Chr.2: 96269306-96269306 on Build GRCh38

View Full Product Details