Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152189775-152190000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA3
Entrez Gene ID
2556
Gene Name
gamma-aminobutyric acid type A receptor alpha3 subunit
Gene Aliases
-
UniGene
Hs.123024
Gene Chromosome Location
Chr.X: 152166234-152451359 on Build GRCh38
Transcripts
NM_000808.3 XM_006724811.2

SNP Detail

SNP ID Chromosome Location
rs767004850 Chr.X: 152189834-152189834 on Build GRCh38
rs1038004936 Chr.X: 152189885-152189885 on Build GRCh38
rs868799149 Chr.X: 152189803-152189803 on Build GRCh38
rs139179813 Chr.X: 152189843-152189843 on Build GRCh38
rs79160441 Chr.X: 152189875-152189875 on Build GRCh38
rs2229939 Chr.X: 152189847-152189847 on Build GRCh38
rs752032476 Chr.X: 152189820-152189820 on Build GRCh38
rs200865926 Chr.X: 152189839-152189839 on Build GRCh38
rs941267146 Chr.X: 152189844-152189844 on Build GRCh38
rs900558124 Chr.X: 152189871-152189871 on Build GRCh38
rs200711028 Chr.X: 152189865-152189865 on Build GRCh38
rs748046661 Chr.X: 152189968-152189968 on Build GRCh38
rs757021133 Chr.X: 152189833-152189833 on Build GRCh38
rs754786466 Chr.X: 152189869-152189869 on Build GRCh38
rs779557844 Chr.X: 152189850-152189850 on Build GRCh38
rs200052640 Chr.X: 152189816-152189816 on Build GRCh38
rs755862388 Chr.X: 152189845-152189845 on Build GRCh38
rs372111492 Chr.X: 152189950-152189950 on Build GRCh38
rs74383358 Chr.X: 152189938-152189938 on Build GRCh38

View Full Product Details