Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125460283-125460540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ1
Entrez Gene ID
9638
Gene Name
fasciculation and elongation protein zeta 1
Gene Aliases
UNC-76
UniGene
Hs.224008
Gene Chromosome Location
Chr.11: 125445745-125496310 on Build GRCh38
Transcripts
NM_005103.4 XM_005271734.2 XM_005271735.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4019037 c.660C>A p.S220S Chr.11: 125460505-125460505 on Build GRCh38
4684379 c.666_667insA p.G223fs*9 Chr.11: 125460498-125460499 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753848285 Chr.11: 125460447-125460447 on Build GRCh38
rs925246921 Chr.11: 125460437-125460437 on Build GRCh38
rs192125986 Chr.11: 125460346-125460346 on Build GRCh38
rs906357039 Chr.11: 125460414-125460414 on Build GRCh38
rs199872494 Chr.11: 125460451-125460451 on Build GRCh38
rs771505558 Chr.11: 125460497-125460497 on Build GRCh38
rs1042404919 Chr.11: 125460428-125460428 on Build GRCh38
rs376391892 Chr.11: 125460471-125460471 on Build GRCh38
rs201547569 Chr.11: 125460470-125460470 on Build GRCh38
rs954228692 Chr.11: 125460347-125460347 on Build GRCh38
rs771689586 Chr.11: 125460515-125460515 on Build GRCh38
rs912838398 Chr.11: 125460409-125460409 on Build GRCh38
rs1045594593 Chr.11: 125460385-125460385 on Build GRCh38
rs987730375 Chr.11: 125460334-125460334 on Build GRCh38
rs778568238 Chr.11: 125460503-125460503 on Build GRCh38
rs745432120 Chr.11: 125460510-125460510 on Build GRCh38
rs987362409 Chr.11: 125460379-125460379 on Build GRCh38
rs750826763 Chr.11: 125460423-125460423 on Build GRCh38
rs142932715 Chr.11: 125460425-125460425 on Build GRCh38
rs532807109 Chr.11: 125460492-125460492 on Build GRCh38
rs544729802 Chr.11: 125460318-125460318 on Build GRCh38
rs1053801703 Chr.11: 125460356-125460356 on Build GRCh38
rs756541178 Chr.11: 125460480-125460480 on Build GRCh38
rs201425669 Chr.11: 125460452-125460452 on Build GRCh38
rs775252980 Chr.11: 125460519-125460519 on Build GRCh38
rs758833210 Chr.11: 125460512-125460512 on Build GRCh38
rs199541284 Chr.11: 125460483-125460483 on Build GRCh38
rs948119353 Chr.11: 125460380-125460380 on Build GRCh38
rs1046956999 Chr.11: 125460442-125460442 on Build GRCh38
rs763365537 Chr.11: 125460359-125460359 on Build GRCh38
rs996602105 Chr.11: 125460514-125460514 on Build GRCh38
rs935296851 Chr.11: 125460349-125460349 on Build GRCh38
rs915292758 Chr.11: 125460358-125460358 on Build GRCh38

View Full Product Details