Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70997988-70998249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCK
Entrez Gene ID
1633
Gene Name
deoxycytidine kinase
Gene Aliases
-
UniGene
Hs.709
Gene Chromosome Location
Chr.4: 70993548-71030914 on Build GRCh38
Transcripts
NM_000788.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640797 c.102G>T p.K34N Chr.4: 70998077-70998077 on Build GRCh38
5799436 c.95C>T p.A32V Chr.4: 70998070-70998070 on Build GRCh38
6640798 c.115A>G p.N39D Chr.4: 70998090-70998090 on Build GRCh38
6276605 c.132G>T p.L44F Chr.4: 70998107-70998107 on Build GRCh38
3727056 c.155C>T p.P52L Chr.4: 70998130-70998130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780459022 Chr.4: 70998044-70998044 on Build GRCh38
rs770521247 Chr.4: 70998094-70998094 on Build GRCh38
rs944159528 Chr.4: 70998095-70998095 on Build GRCh38
rs775124185 Chr.4: 70998129-70998129 on Build GRCh38
rs758981631 Chr.4: 70998104-70998104 on Build GRCh38
rs547107957 Chr.4: 70998154-70998154 on Build GRCh38
rs200062418 Chr.4: 70998031-70998031 on Build GRCh38
rs747405291 Chr.4: 70998090-70998090 on Build GRCh38
rs778365708 Chr.4: 70998214-70998214 on Build GRCh38
rs147385366 Chr.4: 70998017-70998017 on Build GRCh38
rs755124870 Chr.4: 70998055-70998055 on Build GRCh38
rs769291983 Chr.4: 70998123-70998123 on Build GRCh38
rs764136160 Chr.4: 70998151-70998151 on Build GRCh38
rs779113446 Chr.4: 70998063-70998063 on Build GRCh38
rs566429460 Chr.4: 70998138-70998138 on Build GRCh38
rs936222730 Chr.4: 70998149-70998149 on Build GRCh38
rs747313718 Chr.4: 70998215-70998215 on Build GRCh38
rs991159559 Chr.4: 70998028-70998028 on Build GRCh38
rs950365724 Chr.4: 70998050-70998050 on Build GRCh38
rs775494398 Chr.4: 70998200-70998209 on Build GRCh38
rs72552066 Chr.4: 70998204-70998204 on Build GRCh38
rs925604511 Chr.4: 70998053-70998053 on Build GRCh38
rs917476465 Chr.4: 70998032-70998032 on Build GRCh38
rs138544914 Chr.4: 70998153-70998153 on Build GRCh38
rs754105726 Chr.4: 70998196-70998196 on Build GRCh38
rs776277845 Chr.4: 70998100-70998100 on Build GRCh38
rs574859386 Chr.4: 70998172-70998172 on Build GRCh38
rs761726068 Chr.4: 70998166-70998166 on Build GRCh38
rs369907438 Chr.4: 70998206-70998206 on Build GRCh38
rs924760736 Chr.4: 70998116-70998116 on Build GRCh38
rs752858119 Chr.4: 70998212-70998212 on Build GRCh38
rs997498784 Chr.4: 70998168-70998168 on Build GRCh38
rs753922785 Chr.4: 70998045-70998045 on Build GRCh38
rs748587788 Chr.4: 70998064-70998064 on Build GRCh38
rs777965673 Chr.4: 70998087-70998087 on Build GRCh38
rs751420832 Chr.4: 70998017-70998017 on Build GRCh38
rs560907490 Chr.4: 70998134-70998134 on Build GRCh38
rs373489191 Chr.4: 70998071-70998071 on Build GRCh38

View Full Product Details