Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235335164-235335431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGPS1
Entrez Gene ID
9453
Gene Name
geranylgeranyl diphosphate synthase 1
Gene Aliases
GGPPS, GGPPS1
UniGene
Hs.730768
Gene Chromosome Location
Chr.1: 235328438-235344532 on Build GRCh38
Transcripts
NM_001037277.1 NR_036605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4516973 c.24_25CC>TA p.Q9K Chr.1: 235335288-235335289 on Build GRCh38
3689445 c.25C>T p.Q9* Chr.1: 235335289-235335289 on Build GRCh38
905827 c.29G>T p.R10I Chr.1: 235335293-235335293 on Build GRCh38
1253208 c.54C>T p.Y18Y Chr.1: 235335318-235335318 on Build GRCh38
1748236 c.16G>A p.E6K Chr.1: 235335280-235335280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376480594 Chr.1: 235335332-235335332 on Build GRCh38
rs767426473 Chr.1: 235335284-235335284 on Build GRCh38
rs767799955 Chr.1: 235335386-235335386 on Build GRCh38
rs533520208 Chr.1: 235335385-235335385 on Build GRCh38
rs373688757 Chr.1: 235335358-235335358 on Build GRCh38
rs762709232 Chr.1: 235335248-235335248 on Build GRCh38
rs763933452 Chr.1: 235335316-235335319 on Build GRCh38
rs376621224 Chr.1: 235335242-235335242 on Build GRCh38
rs776940011 Chr.1: 235335191-235335191 on Build GRCh38
rs757078597 Chr.1: 235335344-235335344 on Build GRCh38
rs541686720 Chr.1: 235335366-235335366 on Build GRCh38
rs1003158116 Chr.1: 235335203-235335203 on Build GRCh38
rs922131779 Chr.1: 235335394-235335394 on Build GRCh38
rs759817525 Chr.1: 235335268-235335268 on Build GRCh38
rs747726148 Chr.1: 235335362-235335362 on Build GRCh38
rs543784746 Chr.1: 235335340-235335340 on Build GRCh38
rs932110104 Chr.1: 235335406-235335406 on Build GRCh38
rs138463872 Chr.1: 235335354-235335354 on Build GRCh38
rs773181971 Chr.1: 235335382-235335382 on Build GRCh38
rs553538395 Chr.1: 235335211-235335211 on Build GRCh38
rs372026861 Chr.1: 235335218-235335218 on Build GRCh38
rs558003753 Chr.1: 235335273-235335273 on Build GRCh38
rs756984961 Chr.1: 235335221-235335222 on Build GRCh38
rs995074534 Chr.1: 235335190-235335190 on Build GRCh38
rs770005956 Chr.1: 235335350-235335350 on Build GRCh38
rs753701832 Chr.1: 235335316-235335317 on Build GRCh38
rs762938974 Chr.1: 235335383-235335383 on Build GRCh38
rs189645481 Chr.1: 235335262-235335262 on Build GRCh38
rs756527310 Chr.1: 235335302-235335302 on Build GRCh38
rs752833128 Chr.1: 235335293-235335293 on Build GRCh38
rs141445197 Chr.1: 235335357-235335357 on Build GRCh38
rs769794626 Chr.1: 235335237-235335237 on Build GRCh38
rs776210182 Chr.1: 235335222-235335222 on Build GRCh38
rs576374601 Chr.1: 235335319-235335319 on Build GRCh38
rs761599178 Chr.1: 235335230-235335230 on Build GRCh38
rs780520761 Chr.1: 235335360-235335360 on Build GRCh38
rs932875493 Chr.1: 235335401-235335401 on Build GRCh38
rs551812048 Chr.1: 235335392-235335392 on Build GRCh38
rs559999785 Chr.1: 235335384-235335384 on Build GRCh38
rs751772310 Chr.1: 235335266-235335266 on Build GRCh38
rs374596005 Chr.1: 235335318-235335318 on Build GRCh38

View Full Product Details