Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207506631-207506892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR1
Entrez Gene ID
1378
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
Gene Aliases
C3BR, C4BR, CD35, KN
UniGene
Hs.334019
Gene Chromosome Location
Chr.1: 207496128-207641765 on Build GRCh38
Transcripts
NM_000573.3 NM_000651.4 XM_006711166.2 XM_011509205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482659 c.326A>T p.D109V Chr.1: 207506738-207506738 on Build GRCh38
396727 c.327T>A p.D109E Chr.1: 207506739-207506739 on Build GRCh38
3482664 c.333G>A p.V111V Chr.1: 207506745-207506745 on Build GRCh38
4187051 c.367_368delTT p.F123fs*5 Chr.1: 207506779-207506780 on Build GRCh38
5415213 c.313C>T p.R105C Chr.1: 207506725-207506725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769068818 Chr.1: 207506666-207506666 on Build GRCh38
rs763635626 Chr.1: 207506775-207506775 on Build GRCh38
rs745592804 Chr.1: 207506732-207506732 on Build GRCh38
rs774718713 Chr.1: 207506671-207506671 on Build GRCh38
rs376045736 Chr.1: 207506765-207506765 on Build GRCh38
rs187655279 Chr.1: 207506819-207506819 on Build GRCh38
rs527773973 Chr.1: 207506838-207506838 on Build GRCh38
rs370425971 Chr.1: 207506740-207506740 on Build GRCh38
rs574321078 Chr.1: 207506751-207506751 on Build GRCh38
rs755936644 Chr.1: 207506800-207506800 on Build GRCh38
rs11587944 Chr.1: 207506725-207506725 on Build GRCh38
rs765604926 Chr.1: 207506827-207506827 on Build GRCh38
rs751270648 Chr.1: 207506854-207506854 on Build GRCh38
rs55906048 Chr.1: 207506806-207506806 on Build GRCh38
rs565403557 Chr.1: 207506841-207506841 on Build GRCh38
rs750358721 Chr.1: 207506866-207506866 on Build GRCh38
rs769726347 Chr.1: 207506735-207506735 on Build GRCh38
rs761895108 Chr.1: 207506672-207506672 on Build GRCh38
rs183171969 Chr.1: 207506809-207506809 on Build GRCh38
rs540275340 Chr.1: 207506690-207506690 on Build GRCh38
rs750907156 Chr.1: 207506682-207506682 on Build GRCh38
rs377658670 Chr.1: 207506738-207506738 on Build GRCh38
rs3991747 Chr.1: 207506756-207506756 on Build GRCh38
rs878970720 Chr.1: 207506859-207506859 on Build GRCh38
rs752354880 Chr.1: 207506771-207506771 on Build GRCh38
rs777178887 Chr.1: 207506715-207506715 on Build GRCh38
rs767170143 Chr.1: 207506860-207506860 on Build GRCh38
rs3991748 Chr.1: 207506757-207506757 on Build GRCh38
rs748382610 Chr.1: 207506802-207506802 on Build GRCh38
rs757875786 Chr.1: 207506711-207506711 on Build GRCh38
rs773217946 Chr.1: 207506744-207506744 on Build GRCh38
rs554133188 Chr.1: 207506726-207506726 on Build GRCh38
rs571731228 Chr.1: 207506699-207506699 on Build GRCh38
rs368340302 Chr.1: 207506846-207506846 on Build GRCh38
rs563104312 Chr.1: 207506781-207506781 on Build GRCh38
rs756779746 Chr.1: 207506856-207506856 on Build GRCh38
rs964384441 Chr.1: 207506777-207506777 on Build GRCh38
rs760066864 Chr.1: 207506826-207506826 on Build GRCh38
rs751326538 Chr.1: 207506689-207506689 on Build GRCh38
rs761154473 Chr.1: 207506755-207506755 on Build GRCh38
rs771007698 Chr.1: 207506814-207506814 on Build GRCh38
rs767211812 Chr.1: 207506812-207506812 on Build GRCh38
rs754343415 Chr.1: 207506760-207506760 on Build GRCh38
rs754255594 Chr.1: 207506685-207506685 on Build GRCh38
rs993321712 Chr.1: 207506762-207506762 on Build GRCh38
rs55715268 Chr.1: 207506768-207506768 on Build GRCh38
rs374189484 Chr.1: 207506834-207506834 on Build GRCh38
rs779758291 Chr.1: 207506801-207506801 on Build GRCh38
rs746933193 Chr.1: 207506718-207506718 on Build GRCh38
rs781217918 Chr.1: 207506786-207506786 on Build GRCh38
rs3958441 Chr.1: 207506689-207506689 on Build GRCh38
rs745823783 Chr.1: 207506790-207506790 on Build GRCh38
rs914070520 Chr.1: 207506822-207506822 on Build GRCh38
rs764772374 Chr.1: 207506656-207506656 on Build GRCh38
rs767520414 Chr.1: 207506677-207506677 on Build GRCh38
rs55962594 Chr.1: 207506782-207506782 on Build GRCh38

View Full Product Details