Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 61772960-61773233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH4
Entrez Gene ID
1002
Gene Name
cadherin 4
Gene Aliases
CAD4, R-CAD, RCAD
UniGene
Hs.473231
Gene Chromosome Location
Chr.20: 61252426-61940617 on Build GRCh38
Transcripts
NM_001252338.2 NM_001252339.2 NM_001794.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
724919 c.519C>G p.I173M Chr.20: 61773125-61773125 on Build GRCh38
362141 c.555C>A p.P185P Chr.20: 61773161-61773161 on Build GRCh38
7275202 c.397-1G>A p.? Chr.20: 61773002-61773002 on Build GRCh38
6160331 c.398C>T p.P133L Chr.20: 61773004-61773004 on Build GRCh38
5528483 c.444G>A p.P148P Chr.20: 61773050-61773050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773504490 Chr.20: 61773119-61773119 on Build GRCh38
rs140797860 Chr.20: 61773020-61773020 on Build GRCh38
rs745720030 Chr.20: 61773106-61773106 on Build GRCh38
rs775323311 Chr.20: 61773112-61773112 on Build GRCh38
rs573572932 Chr.20: 61773159-61773159 on Build GRCh38
rs748210738 Chr.20: 61773113-61773113 on Build GRCh38
rs933345156 Chr.20: 61773088-61773088 on Build GRCh38
rs187067332 Chr.20: 61773022-61773022 on Build GRCh38
rs990299181 Chr.20: 61772984-61772984 on Build GRCh38
rs752039301 Chr.20: 61773058-61773058 on Build GRCh38
rs202223585 Chr.20: 61773125-61773125 on Build GRCh38
rs766859186 Chr.20: 61773055-61773055 on Build GRCh38
rs755606929 Chr.20: 61773082-61773082 on Build GRCh38
rs766768336 Chr.20: 61773136-61773136 on Build GRCh38
rs756889220 Chr.20: 61773214-61773214 on Build GRCh38
rs770731584 Chr.20: 61773024-61773024 on Build GRCh38
rs1022057718 Chr.20: 61773206-61773206 on Build GRCh38
rs201276877 Chr.20: 61773212-61773212 on Build GRCh38
rs778673063 Chr.20: 61773100-61773100 on Build GRCh38
rs752218061 Chr.20: 61773076-61773076 on Build GRCh38
rs768025577 Chr.20: 61773179-61773179 on Build GRCh38
rs34937312 Chr.20: 61773028-61773028 on Build GRCh38
rs753164006 Chr.20: 61773201-61773201 on Build GRCh38
rs370608791 Chr.20: 61773067-61773067 on Build GRCh38
rs769778529 Chr.20: 61773115-61773115 on Build GRCh38
rs777328179 Chr.20: 61772996-61772996 on Build GRCh38
rs774837151 Chr.20: 61773143-61773143 on Build GRCh38
rs1047152871 Chr.20: 61773090-61773090 on Build GRCh38
rs1003829832 Chr.20: 61773152-61773152 on Build GRCh38
rs373350899 Chr.20: 61773203-61773203 on Build GRCh38
rs763267948 Chr.20: 61773128-61773128 on Build GRCh38
rs765459270 Chr.20: 61773047-61773047 on Build GRCh38
rs745810086 Chr.20: 61773031-61773031 on Build GRCh38
rs868093818 Chr.20: 61773166-61773166 on Build GRCh38
rs113453136 Chr.20: 61773209-61773209 on Build GRCh38
rs905346083 Chr.20: 61773130-61773130 on Build GRCh38
rs974895210 Chr.20: 61773033-61773033 on Build GRCh38
rs771968638 Chr.20: 61773043-61773043 on Build GRCh38
rs1035268864 Chr.20: 61773154-61773154 on Build GRCh38
rs1005735675 Chr.20: 61773105-61773105 on Build GRCh38
rs143400273 Chr.20: 61773093-61773093 on Build GRCh38
rs956401840 Chr.20: 61773195-61773195 on Build GRCh38
rs149210026 Chr.20: 61773086-61773086 on Build GRCh38
rs772016353 Chr.20: 61773111-61773111 on Build GRCh38
rs773592062 Chr.20: 61773048-61773048 on Build GRCh38
rs776758212 Chr.20: 61773044-61773044 on Build GRCh38
rs199952226 Chr.20: 61772989-61772989 on Build GRCh38
rs767954523 Chr.20: 61773073-61773073 on Build GRCh38
rs761840290 Chr.20: 61773046-61773046 on Build GRCh38

View Full Product Details