Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 531999-532254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC34
Entrez Gene ID
997
Gene Name
cell division cycle 34
Gene Aliases
E2-CDC34, UBC3, UBCH3, UBE2R1
UniGene
Hs.514997
Gene Chromosome Location
Chr.19: 531720-542088 on Build GRCh38
Transcripts
NM_004359.1 XM_005259690.3 XM_006722952.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4391714 c.123G>A p.W41* Chr.19: 532054-532054 on Build GRCh38
2927730 c.146C>T p.P49L Chr.19: 532077-532077 on Build GRCh38
2927729 c.100G>A p.D34N Chr.19: 532031-532031 on Build GRCh38
7221076 c.135C>T p.I45I Chr.19: 532066-532066 on Build GRCh38
4629399 c.168C>A p.G56G Chr.19: 532099-532099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902903920 Chr.19: 532168-532168 on Build GRCh38
rs778328367 Chr.19: 532062-532062 on Build GRCh38
rs1013118636 Chr.19: 532215-532215 on Build GRCh38
rs775807294 Chr.19: 532111-532111 on Build GRCh38
rs1052252349 Chr.19: 532115-532115 on Build GRCh38
rs767394877 Chr.19: 532042-532042 on Build GRCh38
rs894621966 Chr.19: 532212-532212 on Build GRCh38
rs1039874495 Chr.19: 532018-532018 on Build GRCh38
rs199984661 Chr.19: 532137-532137 on Build GRCh38
rs755819680 Chr.19: 532044-532044 on Build GRCh38
rs748687986 Chr.19: 532051-532051 on Build GRCh38
rs761826244 Chr.19: 532132-532132 on Build GRCh38
rs1056906674 Chr.19: 532189-532189 on Build GRCh38
rs533730436 Chr.19: 532227-532227 on Build GRCh38
rs750297342 Chr.19: 532043-532043 on Build GRCh38
rs908600490 Chr.19: 532015-532015 on Build GRCh38
rs769854214 Chr.19: 532105-532105 on Build GRCh38
rs757530190 Chr.19: 532162-532162 on Build GRCh38
rs914534066 Chr.19: 532108-532108 on Build GRCh38
rs975917752 Chr.19: 532037-532037 on Build GRCh38
rs746240753 Chr.19: 532097-532097 on Build GRCh38
rs769066610 Chr.19: 532073-532075 on Build GRCh38
rs16989771 Chr.19: 532184-532184 on Build GRCh38
rs774340829 Chr.19: 532123-532123 on Build GRCh38
rs754537445 Chr.19: 532057-532057 on Build GRCh38
rs1002270328 Chr.19: 532202-532202 on Build GRCh38
rs148639611 Chr.19: 532034-532034 on Build GRCh38
rs571173625 Chr.19: 532045-532045 on Build GRCh38
rs757298312 Chr.19: 532038-532038 on Build GRCh38
rs762904994 Chr.19: 532120-532120 on Build GRCh38
rs1035517935 Chr.19: 532213-532213 on Build GRCh38
rs374242233 Chr.19: 532146-532146 on Build GRCh38
rs771486070 Chr.19: 532084-532084 on Build GRCh38
rs374369696 Chr.19: 532033-532033 on Build GRCh38
rs764201386 Chr.19: 532122-532122 on Build GRCh38
rs1004141718 Chr.19: 532224-532224 on Build GRCh38
rs1050765719 Chr.19: 532075-532075 on Build GRCh38
rs368575228 Chr.19: 532087-532087 on Build GRCh38
rs569064607 Chr.19: 532176-532176 on Build GRCh38
rs554753992 Chr.19: 532218-532218 on Build GRCh38
rs751549589 Chr.19: 532036-532036 on Build GRCh38
rs749454138 Chr.19: 532029-532030 on Build GRCh38
rs371106154 Chr.19: 532141-532141 on Build GRCh38
rs6507 Chr.19: 532066-532066 on Build GRCh38
rs774841353 Chr.19: 532138-532138 on Build GRCh38
rs952974907 Chr.19: 532221-532221 on Build GRCh38
rs572980331 Chr.19: 532220-532220 on Build GRCh38
rs965431775 Chr.19: 532233-532233 on Build GRCh38

View Full Product Details