Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67163700-67163925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL8
Entrez Gene ID
55336
Gene Name
F-box and leucine rich repeat protein 8
Gene Aliases
FBL8
UniGene
Hs.710714
Gene Chromosome Location
Chr.16: 67159988-67164174 on Build GRCh38
Transcripts
NM_018378.2 NM_001040667.2 NM_001538.3

Gene Symbol
HSF4
Entrez Gene ID
3299
Gene Name
heat shock transcription factor 4
Gene Aliases
CTM, CTRCT5
UniGene
Hs.512156
Gene Chromosome Location
Chr.16: 67163385-67169945 on Build GRCh38
Transcripts
NM_018378.2 NM_001040667.2 NM_001538.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3510925 c.1116C>A p.L372L Chr.16: 67163811-67163811 on Build GRCh38
5363617 c.1086G>A p.T362T Chr.16: 67163781-67163781 on Build GRCh38
6662012 c.1072A>G p.T358A Chr.16: 67163767-67163767 on Build GRCh38
1718850 c.1090G>A p.E364K Chr.16: 67163785-67163785 on Build GRCh38
5706173 c.1037C>A p.A346D Chr.16: 67163732-67163732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147591659 Chr.16: 67163785-67163785 on Build GRCh38
rs980568961 Chr.16: 67163767-67163767 on Build GRCh38
rs200404151 Chr.16: 67163728-67163728 on Build GRCh38
rs536230683 Chr.16: 67163742-67163742 on Build GRCh38
rs913428549 Chr.16: 67163762-67163762 on Build GRCh38
rs993979498 Chr.16: 67163902-67163902 on Build GRCh38
rs751048283 Chr.16: 67163766-67163766 on Build GRCh38
rs757171481 Chr.16: 67163764-67163771 on Build GRCh38
rs199975180 Chr.16: 67163816-67163816 on Build GRCh38
rs746246454 Chr.16: 67163783-67163783 on Build GRCh38
rs372998678 Chr.16: 67163877-67163878 on Build GRCh38
rs775236230 Chr.16: 67163810-67163810 on Build GRCh38
rs549741253 Chr.16: 67163784-67163784 on Build GRCh38
rs765236872 Chr.16: 67163738-67163738 on Build GRCh38
rs780603966 Chr.16: 67163855-67163855 on Build GRCh38
rs756655584 Chr.16: 67163846-67163846 on Build GRCh38
rs372093553 Chr.16: 67163801-67163801 on Build GRCh38
rs756924028 Chr.16: 67163769-67163769 on Build GRCh38
rs752690771 Chr.16: 67163741-67163741 on Build GRCh38
rs762926798 Chr.16: 67163811-67163811 on Build GRCh38
rs965162501 Chr.16: 67163878-67163879 on Build GRCh38
rs759762267 Chr.16: 67163882-67163883 on Build GRCh38
rs1014290681 Chr.16: 67163722-67163722 on Build GRCh38
rs755561084 Chr.16: 67163867-67163867 on Build GRCh38
rs76287029 Chr.16: 67163873-67163874 on Build GRCh38
rs751099681 Chr.16: 67163836-67163836 on Build GRCh38
rs754228472 Chr.16: 67163857-67163857 on Build GRCh38
rs755783940 Chr.16: 67163853-67163853 on Build GRCh38
rs376436717 Chr.16: 67163852-67163852 on Build GRCh38
rs985088359 Chr.16: 67163797-67163797 on Build GRCh38
rs762161864 Chr.16: 67163726-67163726 on Build GRCh38
rs772347492 Chr.16: 67163802-67163802 on Build GRCh38
rs567180699 Chr.16: 67163733-67163733 on Build GRCh38
rs773252872 Chr.16: 67163804-67163804 on Build GRCh38
rs939031626 Chr.16: 67163849-67163849 on Build GRCh38
rs760941368 Chr.16: 67163805-67163805 on Build GRCh38
rs867407761 Chr.16: 67163878-67163878 on Build GRCh38
rs780668062 Chr.16: 67163779-67163779 on Build GRCh38
rs778744719 Chr.16: 67163831-67163832 on Build GRCh38
rs5817616 Chr.16: 67163875-67163876 on Build GRCh38
rs768911631 Chr.16: 67163720-67163720 on Build GRCh38
rs375269685 Chr.16: 67163723-67163723 on Build GRCh38
rs1038106353 Chr.16: 67163859-67163859 on Build GRCh38
rs767211282 Chr.16: 67163848-67163848 on Build GRCh38
rs946205002 Chr.16: 67163768-67163768 on Build GRCh38
rs765009599 Chr.16: 67163807-67163807 on Build GRCh38
rs763634014 Chr.16: 67163745-67163745 on Build GRCh38
rs578096460 Chr.16: 67163870-67163870 on Build GRCh38
rs774618366 Chr.16: 67163789-67163789 on Build GRCh38
rs558390665 Chr.16: 67163840-67163840 on Build GRCh38
rs367859870 Chr.16: 67163731-67163731 on Build GRCh38
rs397745266 Chr.16: 67163881-67163882 on Build GRCh38

View Full Product Details