Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93876552-93876779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTTIP2
Entrez Gene ID
30836
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Gene Aliases
ERBP, FCF2, HSU15552, LPTS-RP2, TdIF2
UniGene
Hs.601998
Gene Chromosome Location
Chr.1: 93869458-93879206 on Build GRCh38
Transcripts
NM_014597.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1344981 c.1195G>A p.D399N Chr.1: 93876740-93876740 on Build GRCh38
4938556 c.1203A>G p.E401E Chr.1: 93876732-93876732 on Build GRCh38
3806143 c.1261G>C p.E421Q Chr.1: 93876674-93876674 on Build GRCh38
4439153 c.1193A>T p.D398V Chr.1: 93876742-93876742 on Build GRCh38
6647908 c.1261G>A p.E421K Chr.1: 93876674-93876674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374128347 Chr.1: 93876700-93876700 on Build GRCh38
rs774125519 Chr.1: 93876723-93876723 on Build GRCh38
rs775722272 Chr.1: 93876748-93876748 on Build GRCh38
rs776242483 Chr.1: 93876598-93876598 on Build GRCh38
rs544151323 Chr.1: 93876698-93876702 on Build GRCh38
rs373958866 Chr.1: 93876724-93876724 on Build GRCh38
rs775487425 Chr.1: 93876605-93876605 on Build GRCh38
rs200375560 Chr.1: 93876647-93876647 on Build GRCh38
rs759541509 Chr.1: 93876709-93876709 on Build GRCh38
rs772517316 Chr.1: 93876681-93876681 on Build GRCh38
rs768361299 Chr.1: 93876718-93876718 on Build GRCh38
rs763661699 Chr.1: 93876611-93876616 on Build GRCh38
rs180694931 Chr.1: 93876651-93876651 on Build GRCh38
rs778149015 Chr.1: 93876657-93876657 on Build GRCh38
rs764134290 Chr.1: 93876640-93876640 on Build GRCh38
rs886519455 Chr.1: 93876608-93876608 on Build GRCh38
rs575126651 Chr.1: 93876665-93876665 on Build GRCh38
rs369848171 Chr.1: 93876704-93876704 on Build GRCh38
rs375923500 Chr.1: 93876652-93876652 on Build GRCh38
rs898663392 Chr.1: 93876667-93876667 on Build GRCh38
rs747502594 Chr.1: 93876658-93876658 on Build GRCh38
rs920898601 Chr.1: 93876586-93876586 on Build GRCh38
rs764926475 Chr.1: 93876601-93876601 on Build GRCh38
rs35650636 Chr.1: 93876646-93876646 on Build GRCh38
rs186054239 Chr.1: 93876714-93876714 on Build GRCh38
rs751433683 Chr.1: 93876644-93876644 on Build GRCh38
rs780863956 Chr.1: 93876687-93876687 on Build GRCh38
rs993999715 Chr.1: 93876587-93876587 on Build GRCh38
rs35556218 Chr.1: 93876686-93876687 on Build GRCh38
rs771497802 Chr.1: 93876675-93876675 on Build GRCh38
rs759580353 Chr.1: 93876735-93876735 on Build GRCh38
rs758668261 Chr.1: 93876654-93876654 on Build GRCh38
rs960911982 Chr.1: 93876695-93876695 on Build GRCh38
rs752884021 Chr.1: 93876746-93876746 on Build GRCh38
rs1019043587 Chr.1: 93876641-93876641 on Build GRCh38
rs765491955 Chr.1: 93876739-93876739 on Build GRCh38
rs753984933 Chr.1: 93876727-93876727 on Build GRCh38
rs759266462 Chr.1: 93876600-93876600 on Build GRCh38
rs761766261 Chr.1: 93876645-93876645 on Build GRCh38
rs753310025 Chr.1: 93876730-93876732 on Build GRCh38
rs376677281 Chr.1: 93876751-93876751 on Build GRCh38

View Full Product Details