Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 13691941-13692194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
UniGene
Hs.212360
Gene Chromosome Location
Chr.5: 13690328-14011829 on Build GRCh38
Transcripts
NM_001369.2 XM_005248262.3 XM_017009177.1 XM_017009178.1 XM_017009179.1 XM_017009185.1 XM_017009186.1 XM_017009188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
375337 c.13843C>T p.R4615C Chr.5: 13692016-13692016 on Build GRCh38
5596602 c.13849G>A p.V4617I Chr.5: 13692010-13692010 on Build GRCh38
4585414 c.13778C>T p.T4593M Chr.5: 13692081-13692081 on Build GRCh38
5598233 c.13840C>T p.L4614F Chr.5: 13692019-13692019 on Build GRCh38
4648839 c.13798G>A p.A4600T Chr.5: 13692061-13692061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749862482 Chr.5: 13692114-13692114 on Build GRCh38
rs770244453 Chr.5: 13692042-13692042 on Build GRCh38
rs778223884 Chr.5: 13691988-13691988 on Build GRCh38
rs759563638 Chr.5: 13692129-13692129 on Build GRCh38
rs201246418 Chr.5: 13691986-13691986 on Build GRCh38
rs752872674 Chr.5: 13692067-13692067 on Build GRCh38
rs749171065 Chr.5: 13691970-13691970 on Build GRCh38
rs756626821 Chr.5: 13692149-13692153 on Build GRCh38
rs758112779 Chr.5: 13692085-13692085 on Build GRCh38
rs777715365 Chr.5: 13691964-13691964 on Build GRCh38
rs375643128 Chr.5: 13692062-13692062 on Build GRCh38
rs771108785 Chr.5: 13692055-13692055 on Build GRCh38
rs1018054486 Chr.5: 13692006-13692006 on Build GRCh38
rs750336729 Chr.5: 13692167-13692167 on Build GRCh38
rs774279588 Chr.5: 13692061-13692061 on Build GRCh38
rs868145795 Chr.5: 13692124-13692124 on Build GRCh38
rs371639785 Chr.5: 13692044-13692044 on Build GRCh38
rs923678039 Chr.5: 13692010-13692010 on Build GRCh38
rs779655002 Chr.5: 13692169-13692169 on Build GRCh38
rs886059934 Chr.5: 13692034-13692034 on Build GRCh38
rs770287375 Chr.5: 13692112-13692112 on Build GRCh38
rs779884218 Chr.5: 13692090-13692090 on Build GRCh38
rs772086715 Chr.5: 13692139-13692139 on Build GRCh38
rs369094478 Chr.5: 13692113-13692113 on Build GRCh38
rs142036266 Chr.5: 13692081-13692081 on Build GRCh38
rs375610185 Chr.5: 13691984-13691984 on Build GRCh38
rs937336930 Chr.5: 13692171-13692171 on Build GRCh38
rs1036559068 Chr.5: 13692148-13692148 on Build GRCh38
rs771806858 Chr.5: 13692082-13692082 on Build GRCh38
rs746612001 Chr.5: 13692109-13692109 on Build GRCh38
rs760823380 Chr.5: 13692146-13692146 on Build GRCh38
rs745602916 Chr.5: 13692011-13692011 on Build GRCh38
rs774527322 Chr.5: 13692123-13692123 on Build GRCh38
rs753564987 Chr.5: 13692154-13692154 on Build GRCh38
rs367709427 Chr.5: 13692084-13692084 on Build GRCh38
rs376239289 Chr.5: 13692164-13692164 on Build GRCh38
rs147583407 Chr.5: 13692022-13692022 on Build GRCh38
rs867446476 Chr.5: 13692050-13692050 on Build GRCh38
rs201764984 Chr.5: 13692152-13692152 on Build GRCh38
rs771613197 Chr.5: 13692016-13692016 on Build GRCh38
rs775698729 Chr.5: 13692142-13692142 on Build GRCh38
rs760482011 Chr.5: 13692079-13692079 on Build GRCh38
rs191662365 Chr.5: 13692052-13692052 on Build GRCh38
rs374974632 Chr.5: 13692095-13692095 on Build GRCh38
rs748494424 Chr.5: 13692032-13692032 on Build GRCh38
rs756808089 Chr.5: 13691973-13691973 on Build GRCh38
rs200555581 Chr.5: 13692080-13692080 on Build GRCh38
rs997531085 Chr.5: 13692155-13692155 on Build GRCh38
rs771318876 Chr.5: 13692121-13692121 on Build GRCh38
rs760428177 Chr.5: 13692106-13692106 on Build GRCh38
rs778096108 Chr.5: 13692009-13692009 on Build GRCh38
rs2166337 Chr.5: 13692170-13692170 on Build GRCh38

View Full Product Details