Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61816646-61816895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FADS1
Entrez Gene ID
3992
Gene Name
fatty acid desaturase 1
Gene Aliases
D5D, FADS6, FADSD5, LLCDL1, TU12
UniGene
Hs.503546
Gene Chromosome Location
Chr.11: 61799625-61817057 on Build GRCh38
Transcripts
NM_013402.4 NM_001281501.1 NM_001281502.1

Gene Symbol
FADS2
Entrez Gene ID
9415
Gene Name
fatty acid desaturase 2
Gene Aliases
D6D, DES6, FADSD6, LLCDL2, SLL0262, TU13
UniGene
Hs.502745
Gene Chromosome Location
Chr.11: 61816203-61867354 on Build GRCh38
Transcripts
NM_013402.4 NM_001281501.1 NM_001281502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5635033 c.10G>A p.D4N Chr.11: 61816749-61816749 on Build GRCh38
5627753 c.86C>T p.S29L Chr.11: 61816673-61816673 on Build GRCh38
4170344 c.71A>G p.E24G Chr.11: 61816688-61816688 on Build GRCh38
5587756 c.65G>A p.W22* Chr.11: 61816694-61816694 on Build GRCh38
5777789 c.27G>T p.E9D Chr.11: 61816732-61816732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867098916 Chr.11: 61816733-61816733 on Build GRCh38
rs983238070 Chr.11: 61816771-61816771 on Build GRCh38
rs569129600 Chr.11: 61816841-61816841 on Build GRCh38
rs951415436 Chr.11: 61816767-61816767 on Build GRCh38
rs755788898 Chr.11: 61816749-61816749 on Build GRCh38
rs1020577763 Chr.11: 61816786-61816786 on Build GRCh38
rs113766438 Chr.11: 61816855-61816855 on Build GRCh38
rs747092937 Chr.11: 61816876-61816876 on Build GRCh38
rs905098617 Chr.11: 61816694-61816694 on Build GRCh38
rs765576634 Chr.11: 61816689-61816689 on Build GRCh38
rs758868643 Chr.11: 61816725-61816725 on Build GRCh38
rs929374648 Chr.11: 61816869-61816869 on Build GRCh38
rs113305638 Chr.11: 61816858-61816858 on Build GRCh38
rs749484673 Chr.11: 61816738-61816784 on Build GRCh38
rs749035931 Chr.11: 61816783-61816783 on Build GRCh38
rs555414942 Chr.11: 61816814-61816814 on Build GRCh38
rs749976161 Chr.11: 61816732-61816732 on Build GRCh38
rs909222654 Chr.11: 61816877-61816877 on Build GRCh38
rs748090471 Chr.11: 61816803-61816803 on Build GRCh38
rs1019708501 Chr.11: 61816778-61816778 on Build GRCh38
rs979020168 Chr.11: 61816816-61816816 on Build GRCh38
rs753242126 Chr.11: 61816715-61816715 on Build GRCh38
rs11823265 Chr.11: 61816868-61816868 on Build GRCh38
rs995457238 Chr.11: 61816734-61816734 on Build GRCh38
rs773233291 Chr.11: 61816847-61816847 on Build GRCh38
rs779857785 Chr.11: 61816780-61816780 on Build GRCh38
rs965904915 Chr.11: 61816782-61816782 on Build GRCh38
rs910436137 Chr.11: 61816807-61816807 on Build GRCh38
rs941982773 Chr.11: 61816877-61816877 on Build GRCh38
rs921458805 Chr.11: 61816810-61816810 on Build GRCh38
rs11548150 Chr.11: 61816728-61816728 on Build GRCh38
rs771921780 Chr.11: 61816804-61816804 on Build GRCh38
rs979330846 Chr.11: 61816805-61816805 on Build GRCh38
rs935027235 Chr.11: 61816865-61816865 on Build GRCh38
rs759905383 Chr.11: 61816675-61816675 on Build GRCh38
rs535388755 Chr.11: 61816793-61816793 on Build GRCh38
rs976308184 Chr.11: 61816839-61816839 on Build GRCh38

View Full Product Details