Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99903721-99903962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB14
Entrez Gene ID
79982
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Gene Aliases
EGNR9427, PRO34683
UniGene
Hs.577426
Gene Chromosome Location
Chr.4: 99896248-99946726 on Build GRCh38
Transcripts
NM_001031723.3 NM_001278310.1 XM_011532262.2 XM_011532263.2 XM_011532265.2 XM_017008628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5040765 c.887T>G p.V296G Chr.4: 99903854-99903854 on Build GRCh38
1594157 c.985C>T p.R329* Chr.4: 99903756-99903756 on Build GRCh38
4120970 c.978T>C p.T326T Chr.4: 99903763-99903763 on Build GRCh38
4002875 c.852G>A p.G284G Chr.4: 99903889-99903889 on Build GRCh38
4120974 c.922T>A p.Y308N Chr.4: 99903819-99903819 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771075864 Chr.4: 99903811-99903811 on Build GRCh38
rs762826239 Chr.4: 99903850-99903850 on Build GRCh38
rs755548419 Chr.4: 99903917-99903917 on Build GRCh38
rs779256174 Chr.4: 99903755-99903755 on Build GRCh38
rs759530758 Chr.4: 99903841-99903841 on Build GRCh38
rs763886190 Chr.4: 99903862-99903862 on Build GRCh38
rs1031197645 Chr.4: 99903803-99903803 on Build GRCh38
rs143056477 Chr.4: 99903889-99903889 on Build GRCh38
rs767072121 Chr.4: 99903904-99903904 on Build GRCh38
rs751188174 Chr.4: 99903879-99903879 on Build GRCh38
rs767768727 Chr.4: 99903907-99903907 on Build GRCh38
rs747099781 Chr.4: 99903787-99903787 on Build GRCh38
rs775491630 Chr.4: 99903843-99903843 on Build GRCh38
rs905787862 Chr.4: 99903859-99903859 on Build GRCh38
rs748510027 Chr.4: 99903764-99903764 on Build GRCh38
rs757385758 Chr.4: 99903768-99903768 on Build GRCh38
rs149086010 Chr.4: 99903784-99903784 on Build GRCh38
rs776524243 Chr.4: 99903823-99903823 on Build GRCh38
rs1044330763 Chr.4: 99903804-99903804 on Build GRCh38

View Full Product Details