Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150742103-150742350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP1-GIMAP5
Entrez Gene ID
100527949
Gene Name
GIMAP1-GIMAP5 readthrough
Gene Aliases
-
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150716557-150743649 on Build GRCh38
Transcripts
NM_001199577.1 NM_001303630.1 NM_018384.4

Gene Symbol
GIMAP5
Entrez Gene ID
55340
Gene Name
GTPase, IMAP family member 5
Gene Aliases
HIMAP3, IAN-5, IAN4, IAN4L1, IAN5, IMAP3, IROD
UniGene
Hs.647079
Gene Chromosome Location
Chr.7: 150737348-150743649 on Build GRCh38
Transcripts
NM_001199577.1 NM_001303630.1 NM_018384.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6883352 c.98T>C p.V33A Chr.7: 150742237-150742237 on Build GRCh38
3374540 c.131G>T p.G44V Chr.7: 150742270-150742270 on Build GRCh38
3636098 c.74C>T p.P25L Chr.7: 150742213-150742213 on Build GRCh38
3923106 c.165C>T p.S55S Chr.7: 150742304-150742304 on Build GRCh38
6548983 c.71C>T p.T24I Chr.7: 150742210-150742210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746381258 Chr.7: 150742269-150742269 on Build GRCh38
rs376150842 Chr.7: 150742172-150742172 on Build GRCh38
rs761418781 Chr.7: 150742207-150742207 on Build GRCh38
rs753633013 Chr.7: 150742211-150742211 on Build GRCh38
rs373678856 Chr.7: 150742254-150742254 on Build GRCh38
rs377716443 Chr.7: 150742274-150742274 on Build GRCh38
rs774076607 Chr.7: 150742282-150742282 on Build GRCh38
rs781482758 Chr.7: 150742268-150742268 on Build GRCh38
rs750389982 Chr.7: 150742216-150742216 on Build GRCh38
rs145457104 Chr.7: 150742293-150742293 on Build GRCh38
rs757368634 Chr.7: 150742265-150742265 on Build GRCh38
rs200912863 Chr.7: 150742273-150742273 on Build GRCh38
rs773703342 Chr.7: 150742201-150742201 on Build GRCh38
rs905960978 Chr.7: 150742144-150742144 on Build GRCh38
rs1017824445 Chr.7: 150742316-150742316 on Build GRCh38
rs764241258 Chr.7: 150742327-150742327 on Build GRCh38
rs760596434 Chr.7: 150742295-150742295 on Build GRCh38
rs766162569 Chr.7: 150742220-150742220 on Build GRCh38
rs762680564 Chr.7: 150742171-150742171 on Build GRCh38
rs753586959 Chr.7: 150742319-150742319 on Build GRCh38
rs768384469 Chr.7: 150742281-150742281 on Build GRCh38
rs369518198 Chr.7: 150742226-150742226 on Build GRCh38
rs771617924 Chr.7: 150742292-150742292 on Build GRCh38
rs760323946 Chr.7: 150742217-150742217 on Build GRCh38
rs767310158 Chr.7: 150742212-150742212 on Build GRCh38
rs997503558 Chr.7: 150742237-150742237 on Build GRCh38
rs149695569 Chr.7: 150742253-150742253 on Build GRCh38
rs758583284 Chr.7: 150742233-150742233 on Build GRCh38
rs761171921 Chr.7: 150742285-150742285 on Build GRCh38
rs897145688 Chr.7: 150742252-150742252 on Build GRCh38
rs1006060936 Chr.7: 150742312-150742312 on Build GRCh38
rs776063822 Chr.7: 150742161-150742161 on Build GRCh38
rs755455687 Chr.7: 150742238-150742238 on Build GRCh38
rs766339283 Chr.7: 150742314-150742314 on Build GRCh38
rs759345601 Chr.7: 150742324-150742324 on Build GRCh38

View Full Product Details