Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92778020-92778281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEA1
Entrez Gene ID
8411
Gene Name
early endosome antigen 1
Gene Aliases
MST105, MSTP105, ZFYVE2
UniGene
Hs.567367
Gene Chromosome Location
Chr.12: 92772509-92975228 on Build GRCh38
Transcripts
NM_003566.3 XM_011538814.2 XM_011538815.2 XM_017020018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1364864 c.3675G>T p.K1225N Chr.12: 92778159-92778159 on Build GRCh38
4575785 c.3813G>A p.T1271T Chr.12: 92778021-92778021 on Build GRCh38
1317957 c.3812C>T p.T1271M Chr.12: 92778022-92778022 on Build GRCh38
6327451 c.3785G>A p.R1262Q Chr.12: 92778049-92778049 on Build GRCh38
7237526 c.3784C>T p.R1262W Chr.12: 92778050-92778050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903723341 Chr.12: 92778236-92778236 on Build GRCh38
rs752843537 Chr.12: 92778190-92778190 on Build GRCh38
rs1001015088 Chr.12: 92778251-92778251 on Build GRCh38
rs748241824 Chr.12: 92778108-92778108 on Build GRCh38
rs781182648 Chr.12: 92778098-92778098 on Build GRCh38
rs781416585 Chr.12: 92778223-92778223 on Build GRCh38
rs543016668 Chr.12: 92778221-92778221 on Build GRCh38
rs753336875 Chr.12: 92778174-92778174 on Build GRCh38
rs199932446 Chr.12: 92778204-92778204 on Build GRCh38
rs34172720 Chr.12: 92778183-92778183 on Build GRCh38
rs774378824 Chr.12: 92778129-92778129 on Build GRCh38
rs868323627 Chr.12: 92778087-92778087 on Build GRCh38
rs759430688 Chr.12: 92778138-92778138 on Build GRCh38
rs747244654 Chr.12: 92778079-92778079 on Build GRCh38
rs77729665 Chr.12: 92778050-92778050 on Build GRCh38
rs748188677 Chr.12: 92778231-92778231 on Build GRCh38
rs141563687 Chr.12: 92778117-92778117 on Build GRCh38
rs758760167 Chr.12: 92778051-92778051 on Build GRCh38
rs770534947 Chr.12: 92778155-92778155 on Build GRCh38
rs530809966 Chr.12: 92778255-92778255 on Build GRCh38
rs750921917 Chr.12: 92778201-92778201 on Build GRCh38
rs201626769 Chr.12: 92778082-92778082 on Build GRCh38
rs753461435 Chr.12: 92778046-92778046 on Build GRCh38
rs779485484 Chr.12: 92778049-92778049 on Build GRCh38
rs535708993 Chr.12: 92778224-92778224 on Build GRCh38
rs1009465664 Chr.12: 92778175-92778175 on Build GRCh38
rs760399283 Chr.12: 92778163-92778163 on Build GRCh38
rs201651838 Chr.12: 92778060-92778060 on Build GRCh38
rs554527142 Chr.12: 92778189-92778189 on Build GRCh38
rs771877280 Chr.12: 92778139-92778139 on Build GRCh38
rs201094587 Chr.12: 92778173-92778173 on Build GRCh38
rs775747036 Chr.12: 92778159-92778163 on Build GRCh38
rs1022219290 Chr.12: 92778137-92778137 on Build GRCh38
rs574377471 Chr.12: 92778211-92778211 on Build GRCh38

View Full Product Details