Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31760537-31760808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC5
Entrez Gene ID
9681
Gene Name
DEP domain containing 5
Gene Aliases
DEP.5, FFEVF
UniGene
Hs.435022
Gene Chromosome Location
Chr.22: 31753951-31907034 on Build GRCh38
Transcripts
NM_001007188.2 NM_001136029.2 NM_001242896.1 NM_001242897.1 NM_014662.4 NR_110988.1 XM_005261862.1 XM_011530557.2 XM_011530558.2 XM_011530559.2 XM_011530560.2 XM_011530561.2 XM_011530562.2 XM_011530563.2 XM_011530565.2 XM_011530568.2 XM_017029109.1 XM_017029110.1 XM_017029111.1 XM_017029112.1 XM_017029113.1 XM_017029114.1 XM_017029115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019406 c.161A>C p.Q54P Chr.22: 31760670-31760670 on Build GRCh38
5019404 c.161A>C p.Q54P Chr.22: 31760670-31760670 on Build GRCh38
7168790 c.151C>G p.L51V Chr.22: 31760660-31760660 on Build GRCh38
5019405 c.161A>C p.Q54P Chr.22: 31760670-31760670 on Build GRCh38
6642565 c.147-9_147-8insT p.? Chr.22: 31760647-31760648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757433183 Chr.22: 31760565-31760565 on Build GRCh38
rs964318369 Chr.22: 31760727-31760727 on Build GRCh38
rs192624055 Chr.22: 31760639-31760639 on Build GRCh38
rs974482970 Chr.22: 31760729-31760729 on Build GRCh38
rs780602183 Chr.22: 31760610-31760611 on Build GRCh38
rs374688670 Chr.22: 31760631-31760631 on Build GRCh38
rs201312113 Chr.22: 31760670-31760670 on Build GRCh38
rs755742285 Chr.22: 31760720-31760720 on Build GRCh38
rs375999617 Chr.22: 31760614-31760614 on Build GRCh38
rs750036882 Chr.22: 31760718-31760718 on Build GRCh38
rs917210383 Chr.22: 31760571-31760571 on Build GRCh38
rs772602364 Chr.22: 31760632-31760632 on Build GRCh38
rs966689579 Chr.22: 31760676-31760676 on Build GRCh38
rs376099179 Chr.22: 31760714-31760714 on Build GRCh38
rs372718805 Chr.22: 31760696-31760696 on Build GRCh38
rs149515471 Chr.22: 31760622-31760622 on Build GRCh38
rs202081676 Chr.22: 31760620-31760620 on Build GRCh38
rs1013384766 Chr.22: 31760635-31760635 on Build GRCh38
rs538844516 Chr.22: 31760562-31760562 on Build GRCh38
rs886039245 Chr.22: 31760703-31760703 on Build GRCh38
rs768301867 Chr.22: 31760748-31760748 on Build GRCh38
rs756968646 Chr.22: 31760615-31760615 on Build GRCh38
rs922985707 Chr.22: 31760757-31760757 on Build GRCh38
rs369249165 Chr.22: 31760634-31760634 on Build GRCh38
rs765562184 Chr.22: 31760724-31760724 on Build GRCh38
rs755268567 Chr.22: 31760621-31760621 on Build GRCh38
rs774229511 Chr.22: 31760768-31760768 on Build GRCh38
rs184825947 Chr.22: 31760778-31760778 on Build GRCh38
rs1048216242 Chr.22: 31760625-31760625 on Build GRCh38
rs761254536 Chr.22: 31760713-31760713 on Build GRCh38
rs780807875 Chr.22: 31760619-31760619 on Build GRCh38
rs1028446897 Chr.22: 31760755-31760755 on Build GRCh38
rs933499245 Chr.22: 31760758-31760758 on Build GRCh38
rs762835700 Chr.22: 31760674-31760674 on Build GRCh38
rs187819103 Chr.22: 31760610-31760610 on Build GRCh38
rs138015376 Chr.22: 31760600-31760600 on Build GRCh38
rs776930253 Chr.22: 31760659-31760659 on Build GRCh38
rs769423746 Chr.22: 31760672-31760672 on Build GRCh38
rs550525739 Chr.22: 31760689-31760689 on Build GRCh38

View Full Product Details