Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94394933-94395194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL1A2
Entrez Gene ID
1278
Gene Name
collagen type I alpha 2 chain
Gene Aliases
OI4
UniGene
Hs.489142
Gene Chromosome Location
Chr.7: 94394561-94431232 on Build GRCh38
Transcripts
NM_000089.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6406657 c.40G>C p.A14P Chr.7: 94395071-94395071 on Build GRCh38
5813375 c.70+2T>A p.? Chr.7: 94395103-94395103 on Build GRCh38
7298361 c.44T>A p.V15E Chr.7: 94395075-94395075 on Build GRCh38
3642450 c.54C>T p.C18C Chr.7: 94395085-94395085 on Build GRCh38
6111372 c.22C>A p.R8R Chr.7: 94395053-94395053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1042135919 Chr.7: 94394957-94394957 on Build GRCh38
rs762106932 Chr.7: 94395043-94395043 on Build GRCh38
rs777426777 Chr.7: 94395089-94395089 on Build GRCh38
rs770754442 Chr.7: 94395100-94395100 on Build GRCh38
rs1015599975 Chr.7: 94395120-94395120 on Build GRCh38
rs770969589 Chr.7: 94395013-94395013 on Build GRCh38
rs768796844 Chr.7: 94395125-94395125 on Build GRCh38
rs755856571 Chr.7: 94395085-94395085 on Build GRCh38
rs779022418 Chr.7: 94395091-94395091 on Build GRCh38
rs182878866 Chr.7: 94395164-94395164 on Build GRCh38
rs765481774 Chr.7: 94395010-94395010 on Build GRCh38
rs1002844097 Chr.7: 94395115-94395115 on Build GRCh38
rs773423015 Chr.7: 94395159-94395159 on Build GRCh38
rs982206007 Chr.7: 94395109-94395109 on Build GRCh38
rs371620598 Chr.7: 94395152-94395152 on Build GRCh38
rs763326494 Chr.7: 94395160-94395160 on Build GRCh38
rs200278401 Chr.7: 94395083-94395083 on Build GRCh38
rs543810166 Chr.7: 94395051-94395051 on Build GRCh38
rs752079706 Chr.7: 94395078-94395078 on Build GRCh38
rs745803526 Chr.7: 94395124-94395124 on Build GRCh38
rs775681517 Chr.7: 94395015-94395015 on Build GRCh38
rs772606632 Chr.7: 94395151-94395151 on Build GRCh38
rs988694359 Chr.7: 94395072-94395073 on Build GRCh38
rs780687409 Chr.7: 94395079-94395079 on Build GRCh38
rs773394670 Chr.7: 94395123-94395123 on Build GRCh38
rs760889533 Chr.7: 94395017-94395017 on Build GRCh38
rs971349366 Chr.7: 94395086-94395086 on Build GRCh38
rs778946547 Chr.7: 94395123-94395123 on Build GRCh38
rs776925896 Chr.7: 94395027-94395027 on Build GRCh38
rs769411683 Chr.7: 94394987-94394987 on Build GRCh38
rs192298378 Chr.7: 94395097-94395097 on Build GRCh38
rs749148117 Chr.7: 94395093-94395093 on Build GRCh38
rs758795377 Chr.7: 94395060-94395060 on Build GRCh38
rs902276955 Chr.7: 94394972-94394972 on Build GRCh38
rs766918040 Chr.7: 94395065-94395065 on Build GRCh38
rs1032885106 Chr.7: 94394974-94394975 on Build GRCh38
rs761267334 Chr.7: 94395165-94395168 on Build GRCh38
rs371098175 Chr.7: 94395131-94395132 on Build GRCh38
rs766758539 Chr.7: 94395162-94395162 on Build GRCh38
rs551125325 Chr.7: 94395156-94395156 on Build GRCh38
rs998985050 Chr.7: 94395006-94395006 on Build GRCh38
rs764293090 Chr.7: 94395023-94395023 on Build GRCh38
rs773077627 Chr.7: 94394998-94394998 on Build GRCh38

View Full Product Details