Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180313792-180314063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
UniGene
Hs.696497
Gene Chromosome Location
Chr.5: 180300690-180353387 on Build GRCh38
Transcripts
NM_005110.3 XM_006714942.2 XM_017010100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
290168 c.1417G>C p.V473L Chr.5: 180313821-180313821 on Build GRCh38
449465 c.1411A>G p.I471V Chr.5: 180313827-180313827 on Build GRCh38
1238975 c.1370G>A p.R457H Chr.5: 180313868-180313868 on Build GRCh38
6404572 c.1369C>G p.R457G Chr.5: 180313869-180313869 on Build GRCh38
6857039 c.1298C>T p.A433V Chr.5: 180313940-180313940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984145521 Chr.5: 180313979-180313979 on Build GRCh38
rs948578658 Chr.5: 180314040-180314041 on Build GRCh38
rs148353512 Chr.5: 180313875-180313875 on Build GRCh38
rs756896476 Chr.5: 180313846-180313846 on Build GRCh38
rs776590822 Chr.5: 180313995-180313995 on Build GRCh38
rs368152280 Chr.5: 180313954-180313954 on Build GRCh38
rs980615089 Chr.5: 180313885-180313885 on Build GRCh38
rs201795453 Chr.5: 180313949-180313949 on Build GRCh38
rs774769824 Chr.5: 180313975-180313975 on Build GRCh38
rs750371085 Chr.5: 180314001-180314001 on Build GRCh38
rs763077671 Chr.5: 180313810-180313810 on Build GRCh38
rs977514512 Chr.5: 180313851-180313851 on Build GRCh38
rs753867862 Chr.5: 180313915-180313915 on Build GRCh38
rs895891209 Chr.5: 180314033-180314033 on Build GRCh38
rs759729689 Chr.5: 180313820-180313820 on Build GRCh38
rs760448629 Chr.5: 180313906-180313906 on Build GRCh38
rs745566290 Chr.5: 180313860-180313860 on Build GRCh38
rs757240985 Chr.5: 180313918-180313918 on Build GRCh38
rs771044452 Chr.5: 180313881-180313881 on Build GRCh38
rs1044617199 Chr.5: 180314041-180314041 on Build GRCh38
rs377416267 Chr.5: 180313891-180313891 on Build GRCh38
rs897463241 Chr.5: 180313955-180313955 on Build GRCh38
rs1012796939 Chr.5: 180313899-180313899 on Build GRCh38
rs748473714 Chr.5: 180313867-180313867 on Build GRCh38
rs987196613 Chr.5: 180313928-180313928 on Build GRCh38
rs902054261 Chr.5: 180313994-180313994 on Build GRCh38
rs779630983 Chr.5: 180313936-180313936 on Build GRCh38
rs374321706 Chr.5: 180313982-180313982 on Build GRCh38
rs777589155 Chr.5: 180313843-180313843 on Build GRCh38
rs745952615 Chr.5: 180313978-180313978 on Build GRCh38
rs761660822 Chr.5: 180313997-180313997 on Build GRCh38
rs768500736 Chr.5: 180314037-180314037 on Build GRCh38
rs778086543 Chr.5: 180313945-180313945 on Build GRCh38
rs756474057 Chr.5: 180313939-180313939 on Build GRCh38
rs764031517 Chr.5: 180313988-180313988 on Build GRCh38
rs930300666 Chr.5: 180313977-180313977 on Build GRCh38
rs774433246 Chr.5: 180313888-180313888 on Build GRCh38
rs766439482 Chr.5: 180313817-180313817 on Build GRCh38
rs975563453 Chr.5: 180313957-180313957 on Build GRCh38
rs749984616 Chr.5: 180313930-180313930 on Build GRCh38
rs889245305 Chr.5: 180313816-180313816 on Build GRCh38
rs532880612 Chr.5: 180313831-180313831 on Build GRCh38
rs746667820 Chr.5: 180313938-180313938 on Build GRCh38
rs759042416 Chr.5: 180313881-180313882 on Build GRCh38
rs370443163 Chr.5: 180313920-180313920 on Build GRCh38
rs778365839 Chr.5: 180313962-180313968 on Build GRCh38
rs765218880 Chr.5: 180313919-180313919 on Build GRCh38
rs775727574 Chr.5: 180313900-180313900 on Build GRCh38
rs1057112747 Chr.5: 180314000-180314000 on Build GRCh38
rs753816564 Chr.5: 180313844-180313844 on Build GRCh38
rs377307097 Chr.5: 180313861-180313861 on Build GRCh38
rs947850125 Chr.5: 180313879-180313879 on Build GRCh38
rs767726394 Chr.5: 180313821-180313821 on Build GRCh38
rs199809518 Chr.5: 180313849-180313849 on Build GRCh38
rs141497441 Chr.5: 180313980-180313980 on Build GRCh38
rs571223936 Chr.5: 180313948-180313948 on Build GRCh38
rs562345769 Chr.5: 180313848-180313848 on Build GRCh38
rs763947287 Chr.5: 180313907-180313907 on Build GRCh38
rs765278379 Chr.5: 180313998-180313998 on Build GRCh38
rs759616421 Chr.5: 180313889-180313889 on Build GRCh38
rs371138776 Chr.5: 180313869-180313869 on Build GRCh38
rs931551413 Chr.5: 180313969-180313969 on Build GRCh38
rs2303007 Chr.5: 180313827-180313827 on Build GRCh38
rs915178519 Chr.5: 180313878-180313878 on Build GRCh38
rs759975229 Chr.5: 180313963-180313963 on Build GRCh38
rs757867551 Chr.5: 180313935-180313935 on Build GRCh38

View Full Product Details