Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157321567-157321826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERMN
Entrez Gene ID
57471
Gene Name
ermin
Gene Aliases
JN, KIAA1189
UniGene
Hs.443894
Gene Chromosome Location
Chr.2: 157318613-157327713 on Build GRCh38
Transcripts
NM_001009959.2 NM_001304344.1 NM_001304345.1 NM_001304346.1 NM_020711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251485 c.592G>C p.E198Q Chr.2: 157321573-157321573 on Build GRCh38
4390402 c.548T>C p.M183T Chr.2: 157321617-157321617 on Build GRCh38
6088282 c.483G>A p.K161K Chr.2: 157321682-157321682 on Build GRCh38
6088283 c.546C>T p.D182D Chr.2: 157321619-157321619 on Build GRCh38
5901131 c.452C>T p.P151L Chr.2: 157321713-157321713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749347485 Chr.2: 157321687-157321687 on Build GRCh38
rs773232236 Chr.2: 157321594-157321594 on Build GRCh38
rs771950532 Chr.2: 157321617-157321617 on Build GRCh38
rs781590531 Chr.2: 157321674-157321674 on Build GRCh38
rs183779910 Chr.2: 157321758-157321758 on Build GRCh38
rs372196713 Chr.2: 157321715-157321715 on Build GRCh38
rs777887214 Chr.2: 157321772-157321772 on Build GRCh38
rs763916818 Chr.2: 157321635-157321635 on Build GRCh38
rs778032477 Chr.2: 157321686-157321686 on Build GRCh38
rs376833308 Chr.2: 157321733-157321733 on Build GRCh38
rs776307442 Chr.2: 157321620-157321620 on Build GRCh38
rs765669373 Chr.2: 157321659-157321659 on Build GRCh38
rs757299320 Chr.2: 157321781-157321781 on Build GRCh38
rs926028768 Chr.2: 157321651-157321651 on Build GRCh38
rs771004596 Chr.2: 157321693-157321693 on Build GRCh38
rs764849682 Chr.2: 157321718-157321718 on Build GRCh38
rs781148830 Chr.2: 157321604-157321604 on Build GRCh38
rs1055709186 Chr.2: 157321757-157321757 on Build GRCh38
rs895977284 Chr.2: 157321784-157321784 on Build GRCh38
rs946132094 Chr.2: 157321603-157321603 on Build GRCh38
rs752034292 Chr.2: 157321663-157321663 on Build GRCh38
rs140855571 Chr.2: 157321759-157321759 on Build GRCh38
rs374683748 Chr.2: 157321621-157321621 on Build GRCh38
rs749356792 Chr.2: 157321595-157321595 on Build GRCh38
rs781158950 Chr.2: 157321739-157321741 on Build GRCh38
rs760084401 Chr.2: 157321631-157321631 on Build GRCh38
rs775296669 Chr.2: 157321696-157321696 on Build GRCh38
rs368029065 Chr.2: 157321700-157321700 on Build GRCh38
rs761673159 Chr.2: 157321717-157321717 on Build GRCh38
rs376770964 Chr.2: 157321773-157321773 on Build GRCh38
rs772874329 Chr.2: 157321726-157321726 on Build GRCh38
rs200919525 Chr.2: 157321763-157321763 on Build GRCh38
rs760097906 Chr.2: 157321761-157321761 on Build GRCh38
rs757906549 Chr.2: 157321655-157321655 on Build GRCh38
rs756451218 Chr.2: 157321771-157321771 on Build GRCh38
rs921392312 Chr.2: 157321697-157321697 on Build GRCh38
rs776437441 Chr.2: 157321636-157321636 on Build GRCh38
rs554539666 Chr.2: 157321638-157321638 on Build GRCh38
rs775447600 Chr.2: 157321618-157321618 on Build GRCh38
rs746885969 Chr.2: 157321698-157321698 on Build GRCh38
rs755346464 Chr.2: 157321673-157321673 on Build GRCh38
rs779107610 Chr.2: 157321794-157321794 on Build GRCh38
rs749945115 Chr.2: 157321652-157321652 on Build GRCh38
rs745938470 Chr.2: 157321795-157321795 on Build GRCh38
rs767981317 Chr.2: 157321762-157321762 on Build GRCh38

View Full Product Details