Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46745970-46746219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKRP
Entrez Gene ID
79147
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
UniGene
Hs.515493
Gene Chromosome Location
Chr.19: 46746015-46758575 on Build GRCh38
Transcripts
NM_001039885.2 NM_024301.4 XM_005259247.2 XM_005259248.2 XM_005259249.4 XM_011527306.1 XM_011527307.1 XM_017027297.1 NM_001039877.1 NM_013403.2 XM_006723171.2 XM_006723172.2 XM_011526878.1 XM_017026719.1

Gene Symbol
STRN4
Entrez Gene ID
29888
Gene Name
striatin 4
Gene Aliases
PPP2R6C, ZIN, zinedin
UniGene
Hs.631590
Gene Chromosome Location
Chr.19: 46719509-46746475 on Build GRCh38
Transcripts
NM_001039885.2 NM_024301.4 XM_005259247.2 XM_005259248.2 XM_005259249.4 XM_011527306.1 XM_011527307.1 XM_017027297.1 NM_001039877.1 NM_013403.2 XM_006723171.2 XM_006723172.2 XM_011526878.1 XM_017026719.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6299511 c.273C>A p.A91A Chr.19: 46746158-46746158 on Build GRCh38
6299512 c.273C>A p.A91A Chr.19: 46746158-46746158 on Build GRCh38
5017574 c.213delT p.F71fs*40 Chr.19: 46746218-46746218 on Build GRCh38
5017573 c.213delT p.F71fs*40 Chr.19: 46746218-46746218 on Build GRCh38
4955166 c.250G>T p.A84S Chr.19: 46746181-46746181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003023306 Chr.19: 46746067-46746067 on Build GRCh38
rs978879395 Chr.19: 46746113-46746113 on Build GRCh38
rs568014119 Chr.19: 46746094-46746095 on Build GRCh38
rs762105525 Chr.19: 46746129-46746129 on Build GRCh38
rs540282620 Chr.19: 46745993-46745993 on Build GRCh38
rs535458876 Chr.19: 46746159-46746159 on Build GRCh38
rs867253089 Chr.19: 46746036-46746036 on Build GRCh38
rs767225114 Chr.19: 46746176-46746176 on Build GRCh38
rs779977307 Chr.19: 46746115-46746115 on Build GRCh38
rs566882151 Chr.19: 46746157-46746157 on Build GRCh38
rs531446425 Chr.19: 46746083-46746083 on Build GRCh38
rs764406449 Chr.19: 46746113-46746114 on Build GRCh38
rs924689158 Chr.19: 46746114-46746115 on Build GRCh38
rs546744915 Chr.19: 46746155-46746155 on Build GRCh38
rs1015287271 Chr.19: 46746070-46746071 on Build GRCh38
rs865996066 Chr.19: 46746163-46746163 on Build GRCh38
rs533261221 Chr.19: 46746123-46746123 on Build GRCh38
rs866528952 Chr.19: 46746150-46746150 on Build GRCh38
rs766953974 Chr.19: 46746146-46746146 on Build GRCh38
rs1026658036 Chr.19: 46746095-46746099 on Build GRCh38
rs1023339872 Chr.19: 46746005-46746005 on Build GRCh38
rs763404097 Chr.19: 46746039-46746039 on Build GRCh38
rs12984041 Chr.19: 46746039-46746039 on Build GRCh38
rs776859717 Chr.19: 46746125-46746125 on Build GRCh38
rs571064207 Chr.19: 46746099-46746099 on Build GRCh38
rs201764053 Chr.19: 46746201-46746201 on Build GRCh38
rs763353721 Chr.19: 46746136-46746136 on Build GRCh38
rs770961397 Chr.19: 46746112-46746112 on Build GRCh38
rs965228907 Chr.19: 46746109-46746114 on Build GRCh38
rs551160185 Chr.19: 46746096-46746096 on Build GRCh38
rs749236827 Chr.19: 46746110-46746110 on Build GRCh38
rs770147043 Chr.19: 46746134-46746134 on Build GRCh38
rs995093352 Chr.19: 46746071-46746076 on Build GRCh38
rs777788662 Chr.19: 46746106-46746106 on Build GRCh38
rs1005294505 Chr.19: 46746004-46746004 on Build GRCh38
rs755899946 Chr.19: 46746181-46746181 on Build GRCh38
rs183340182 Chr.19: 46746021-46746021 on Build GRCh38
rs560100018 Chr.19: 46746008-46746008 on Build GRCh38
rs773630493 Chr.19: 46746135-46746135 on Build GRCh38
rs1034659793 Chr.19: 46746037-46746037 on Build GRCh38
rs1019972624 Chr.19: 46746103-46746103 on Build GRCh38
rs985412051 Chr.19: 46746101-46746101 on Build GRCh38
rs912410742 Chr.19: 46746015-46746015 on Build GRCh38
rs934736564 Chr.19: 46746117-46746117 on Build GRCh38
rs752405093 Chr.19: 46746179-46746179 on Build GRCh38
rs1034551152 Chr.19: 46746027-46746027 on Build GRCh38
rs562627844 Chr.19: 46746040-46746040 on Build GRCh38
rs781029124 Chr.19: 46746098-46746098 on Build GRCh38
rs944763479 Chr.19: 46746020-46746020 on Build GRCh38

View Full Product Details