Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141640129-141640384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHSD1
Entrez Gene ID
89848
Gene Name
FCH and double SH3 domains 1
Gene Aliases
NWK2
UniGene
Hs.591257
Gene Chromosome Location
Chr.5: 141639302-141651453 on Build GRCh38
Transcripts
NM_033449.2 NM_001130029.1 NM_173828.4 XM_005268414.3 XM_011537624.1 XM_011537625.2

Gene Symbol
RELL2
Entrez Gene ID
285613
Gene Name
RELT like 2
Gene Aliases
C5orf16
UniGene
Hs.120094
Gene Chromosome Location
Chr.5: 141636946-141641077 on Build GRCh38
Transcripts
NM_033449.2 NM_001130029.1 NM_173828.4 XM_005268414.3 XM_011537624.1 XM_011537625.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1223511 c.727A>G p.R243G Chr.5: 141640143-141640143 on Build GRCh38
5961790 c.754C>T p.L252F Chr.5: 141640170-141640170 on Build GRCh38
4159581 c.847C>G p.Q283E Chr.5: 141640263-141640263 on Build GRCh38
5918800 c.781G>A p.E261K Chr.5: 141640197-141640197 on Build GRCh38
295185 c.862G>A p.D288N Chr.5: 141640278-141640278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748779251 Chr.5: 141640259-141640259 on Build GRCh38
rs769298620 Chr.5: 141640311-141640311 on Build GRCh38
rs760874296 Chr.5: 141640304-141640305 on Build GRCh38
rs764086935 Chr.5: 141640187-141640187 on Build GRCh38
rs767764451 Chr.5: 141640169-141640169 on Build GRCh38
rs369892061 Chr.5: 141640192-141640192 on Build GRCh38
rs543245703 Chr.5: 141640214-141640214 on Build GRCh38
rs772716969 Chr.5: 141640157-141640157 on Build GRCh38
rs532286514 Chr.5: 141640219-141640219 on Build GRCh38
rs748788130 Chr.5: 141640341-141640341 on Build GRCh38
rs35340436 Chr.5: 141640175-141640176 on Build GRCh38
rs758921383 Chr.5: 141640328-141640328 on Build GRCh38
rs369105841 Chr.5: 141640325-141640325 on Build GRCh38
rs750797330 Chr.5: 141640210-141640210 on Build GRCh38
rs761133000 Chr.5: 141640309-141640309 on Build GRCh38
rs200320253 Chr.5: 141640228-141640228 on Build GRCh38
rs771839658 Chr.5: 141640293-141640293 on Build GRCh38
rs924668013 Chr.5: 141640226-141640226 on Build GRCh38
rs762800363 Chr.5: 141640161-141640161 on Build GRCh38
rs367620461 Chr.5: 141640351-141640351 on Build GRCh38
rs762186023 Chr.5: 141640255-141640255 on Build GRCh38
rs199786252 Chr.5: 141640313-141640313 on Build GRCh38
rs747660127 Chr.5: 141640286-141640286 on Build GRCh38
rs752911612 Chr.5: 141640170-141640170 on Build GRCh38
rs769418606 Chr.5: 141640150-141640150 on Build GRCh38
rs760800991 Chr.5: 141640173-141640173 on Build GRCh38
rs372439670 Chr.5: 141640215-141640215 on Build GRCh38
rs774311343 Chr.5: 141640270-141640270 on Build GRCh38
rs1049731746 Chr.5: 141640246-141640246 on Build GRCh38
rs770859414 Chr.5: 141640164-141640164 on Build GRCh38
rs775199549 Chr.5: 141640307-141640307 on Build GRCh38
rs759392634 Chr.5: 141640281-141640281 on Build GRCh38
rs758833121 Chr.5: 141640212-141640212 on Build GRCh38
rs779454867 Chr.5: 141640205-141640205 on Build GRCh38
rs768261083 Chr.5: 141640176-141640176 on Build GRCh38
rs759302837 Chr.5: 141640168-141640168 on Build GRCh38
rs755741628 Chr.5: 141640340-141640340 on Build GRCh38
rs751928779 Chr.5: 141640337-141640337 on Build GRCh38
rs32956 Chr.5: 141640314-141640314 on Build GRCh38
rs11742646 Chr.5: 141640263-141640263 on Build GRCh38
rs569973650 Chr.5: 141640191-141640191 on Build GRCh38
rs769500107 Chr.5: 141640221-141640221 on Build GRCh38
rs936785930 Chr.5: 141640204-141640204 on Build GRCh38
rs377253130 Chr.5: 141640335-141640335 on Build GRCh38
rs764330298 Chr.5: 141640312-141640312 on Build GRCh38
rs934732291 Chr.5: 141640249-141640250 on Build GRCh38
rs375807322 Chr.5: 141640165-141640165 on Build GRCh38

View Full Product Details