Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116860996-116861221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
NM_001242699.1 XM_005269815.3 XM_006717835.3 XM_006717836.3 XM_011539797.2 XM_011539799.2 XM_017016241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5927581 c.869C>T p.S290L Chr.10: 116861120-116861120 on Build GRCh38
6518324 c.925T>A p.L309I Chr.10: 116861176-116861176 on Build GRCh38
2054882 c.821C>T p.T274M Chr.10: 116861072-116861072 on Build GRCh38
5937428 c.821C>G p.T274R Chr.10: 116861072-116861072 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897120022 Chr.10: 116861073-116861073 on Build GRCh38
rs565754588 Chr.10: 116861041-116861041 on Build GRCh38
rs892021844 Chr.10: 116861110-116861110 on Build GRCh38
rs954113919 Chr.10: 116861113-116861113 on Build GRCh38
rs756132878 Chr.10: 116861163-116861163 on Build GRCh38
rs764415269 Chr.10: 116861076-116861076 on Build GRCh38
rs978714805 Chr.10: 116861155-116861155 on Build GRCh38
rs1037044924 Chr.10: 116861056-116861056 on Build GRCh38
rs961214578 Chr.10: 116861046-116861046 on Build GRCh38
rs771733893 Chr.10: 116861031-116861032 on Build GRCh38
rs1005169750 Chr.10: 116861045-116861045 on Build GRCh38
rs1053390256 Chr.10: 116861100-116861100 on Build GRCh38
rs757784533 Chr.10: 116861136-116861136 on Build GRCh38
rs948777733 Chr.10: 116861068-116861068 on Build GRCh38
rs774919854 Chr.10: 116861088-116861088 on Build GRCh38
rs928661821 Chr.10: 116861082-116861082 on Build GRCh38
rs758900693 Chr.10: 116861030-116861030 on Build GRCh38
rs968331175 Chr.10: 116861152-116861152 on Build GRCh38
rs192163696 Chr.10: 116861137-116861137 on Build GRCh38
rs973850332 Chr.10: 116861047-116861047 on Build GRCh38
rs1007044604 Chr.10: 116861129-116861129 on Build GRCh38
rs750508798 Chr.10: 116861161-116861161 on Build GRCh38
rs773386394 Chr.10: 116861139-116861139 on Build GRCh38
rs947897687 Chr.10: 116861031-116861031 on Build GRCh38
rs762287866 Chr.10: 116861097-116861097 on Build GRCh38
rs558716110 Chr.10: 116861035-116861035 on Build GRCh38
rs764958511 Chr.10: 116861036-116861036 on Build GRCh38
rs1048917489 Chr.10: 116861040-116861040 on Build GRCh38
rs1043578597 Chr.10: 116861166-116861166 on Build GRCh38
rs772651228 Chr.10: 116861104-116861104 on Build GRCh38
rs763445380 Chr.10: 116861072-116861072 on Build GRCh38

View Full Product Details