Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63555576-63555833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC11
Entrez Gene ID
79025
Gene Name
fibronectin type III domain containing 11
Gene Aliases
C20orf195
UniGene
Hs.197755
Gene Chromosome Location
Chr.20: 63553020-63556712 on Build GRCh38
Transcripts
NM_001319152.1 NM_001319153.1 NM_024059.3 XM_011529041.2 XM_017028060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615548 c.111G>A p.A37A Chr.20: 63555774-63555774 on Build GRCh38
3963792 c.15G>T p.V5V Chr.20: 63555678-63555678 on Build GRCh38
1578750 c.132C>T p.R44R Chr.20: 63555795-63555795 on Build GRCh38
4888903 c.42G>T p.K14N Chr.20: 63555705-63555705 on Build GRCh38
7194112 c.33C>T p.D11D Chr.20: 63555696-63555696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570697040 Chr.20: 63555713-63555713 on Build GRCh38
rs751712194 Chr.20: 63555668-63555668 on Build GRCh38
rs763240266 Chr.20: 63555619-63555619 on Build GRCh38
rs1043665422 Chr.20: 63555664-63555664 on Build GRCh38
rs764141769 Chr.20: 63555649-63555649 on Build GRCh38
rs767735709 Chr.20: 63555776-63555776 on Build GRCh38
rs745472289 Chr.20: 63555739-63555739 on Build GRCh38
rs767088319 Chr.20: 63555642-63555642 on Build GRCh38
rs374151368 Chr.20: 63555782-63555782 on Build GRCh38
rs768261839 Chr.20: 63555807-63555807 on Build GRCh38
rs140558959 Chr.20: 63555811-63555811 on Build GRCh38
rs530838389 Chr.20: 63555646-63555646 on Build GRCh38
rs753195794 Chr.20: 63555742-63555758 on Build GRCh38
rs552539833 Chr.20: 63555688-63555688 on Build GRCh38
rs779082342 Chr.20: 63555794-63555794 on Build GRCh38
rs939158739 Chr.20: 63555629-63555629 on Build GRCh38
rs775280992 Chr.20: 63555800-63555800 on Build GRCh38
rs376299359 Chr.20: 63555680-63555680 on Build GRCh38
rs761862075 Chr.20: 63555772-63555772 on Build GRCh38
rs765122577 Chr.20: 63555659-63555659 on Build GRCh38
rs763767284 Chr.20: 63555750-63555750 on Build GRCh38
rs771384731 Chr.20: 63555637-63555637 on Build GRCh38
rs199555784 Chr.20: 63555789-63555789 on Build GRCh38
rs369332991 Chr.20: 63555809-63555809 on Build GRCh38
rs773868805 Chr.20: 63555639-63555639 on Build GRCh38
rs765790166 Chr.20: 63555734-63555736 on Build GRCh38
rs148340283 Chr.20: 63555773-63555773 on Build GRCh38
rs776230427 Chr.20: 63555808-63555808 on Build GRCh38
rs749872664 Chr.20: 63555712-63555712 on Build GRCh38
rs563785888 Chr.20: 63555607-63555607 on Build GRCh38
rs773495662 Chr.20: 63555618-63555618 on Build GRCh38
rs1054024063 Chr.20: 63555783-63555783 on Build GRCh38
rs558101947 Chr.20: 63555796-63555796 on Build GRCh38
rs771857316 Chr.20: 63555740-63555740 on Build GRCh38
rs1046152158 Chr.20: 63555713-63555714 on Build GRCh38
rs774825951 Chr.20: 63555734-63555734 on Build GRCh38
rs775082377 Chr.20: 63555741-63555741 on Build GRCh38
rs755123776 Chr.20: 63555775-63555775 on Build GRCh38
rs200397679 Chr.20: 63555647-63555647 on Build GRCh38
rs757814732 Chr.20: 63555793-63555793 on Build GRCh38
rs760249867 Chr.20: 63555744-63555744 on Build GRCh38
rs749735513 Chr.20: 63555790-63555790 on Build GRCh38
rs781206977 Chr.20: 63555686-63555686 on Build GRCh38
rs759272761 Chr.20: 63555641-63555641 on Build GRCh38
rs758448112 Chr.20: 63555793-63555794 on Build GRCh38
rs755211779 Chr.20: 63555669-63555669 on Build GRCh38
rs778239532 Chr.20: 63555690-63555690 on Build GRCh38
rs993610915 Chr.20: 63555795-63555795 on Build GRCh38
rs752973095 Chr.20: 63555779-63555779 on Build GRCh38
rs750260904 Chr.20: 63555774-63555774 on Build GRCh38
rs369567461 Chr.20: 63555771-63555771 on Build GRCh38
rs756552239 Chr.20: 63555689-63555689 on Build GRCh38
rs753874919 Chr.20: 63555650-63555650 on Build GRCh38
rs375992607 Chr.20: 63555797-63555797 on Build GRCh38
rs746703736 Chr.20: 63555806-63555806 on Build GRCh38
rs757003025 Chr.20: 63555658-63555658 on Build GRCh38

View Full Product Details