Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98943993-98944264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL15A1
Entrez Gene ID
1306
Gene Name
collagen type XV alpha 1 chain
Gene Aliases
-
UniGene
Hs.409034
Gene Chromosome Location
Chr.9: 98943392-99070792 on Build GRCh38
Transcripts
NM_001855.4 XM_011518214.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361994 c.47T>C p.L16P Chr.9: 98944197-98944197 on Build GRCh38
3952106 c.62C>A p.T21K Chr.9: 98944212-98944212 on Build GRCh38
1103161 c.67C>A p.L23I Chr.9: 98944217-98944217 on Build GRCh38
4759494 c.89G>A p.R30H Chr.9: 98944239-98944239 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776895783 Chr.9: 98944029-98944029 on Build GRCh38
rs775665306 Chr.9: 98944021-98944021 on Build GRCh38
rs774761012 Chr.9: 98944214-98944214 on Build GRCh38
rs145179644 Chr.9: 98944241-98944241 on Build GRCh38
rs765553416 Chr.9: 98944040-98944040 on Build GRCh38
rs773757219 Chr.9: 98944041-98944041 on Build GRCh38
rs761736585 Chr.9: 98944230-98944230 on Build GRCh38
rs200514853 Chr.9: 98944022-98944022 on Build GRCh38
rs752393916 Chr.9: 98944056-98944056 on Build GRCh38
rs1020932695 Chr.9: 98944238-98944238 on Build GRCh38
rs772331324 Chr.9: 98944020-98944020 on Build GRCh38
rs762147016 Chr.9: 98944034-98944034 on Build GRCh38
rs780194163 Chr.9: 98944082-98944082 on Build GRCh38
rs769617874 Chr.9: 98944013-98944013 on Build GRCh38
rs755780960 Chr.9: 98944147-98944147 on Build GRCh38
rs556824568 Chr.9: 98944109-98944109 on Build GRCh38
rs749210426 Chr.9: 98944018-98944018 on Build GRCh38
rs766815864 Chr.9: 98944116-98944116 on Build GRCh38
rs747112212 Chr.9: 98944088-98944088 on Build GRCh38
rs771506945 Chr.9: 98944216-98944216 on Build GRCh38
rs750969536 Chr.9: 98944051-98944051 on Build GRCh38
rs897482912 Chr.9: 98944084-98944084 on Build GRCh38
rs753626754 Chr.9: 98944152-98944152 on Build GRCh38
rs748399210 Chr.9: 98944180-98944180 on Build GRCh38
rs577588200 Chr.9: 98944225-98944225 on Build GRCh38
rs778778001 Chr.9: 98944167-98944167 on Build GRCh38
rs768700306 Chr.9: 98944024-98944024 on Build GRCh38
rs201993304 Chr.9: 98944172-98944172 on Build GRCh38
rs374798276 Chr.9: 98944199-98944199 on Build GRCh38
rs372448914 Chr.9: 98944081-98944081 on Build GRCh38
rs770165221 Chr.9: 98944105-98944105 on Build GRCh38
rs777929387 Chr.9: 98944015-98944015 on Build GRCh38
rs375160252 Chr.9: 98944146-98944146 on Build GRCh38
rs200458089 Chr.9: 98944012-98944012 on Build GRCh38
rs765102697 Chr.9: 98944098-98944106 on Build GRCh38
rs1048438333 Chr.9: 98944077-98944077 on Build GRCh38
rs540101582 Chr.9: 98944228-98944228 on Build GRCh38
rs757102272 Chr.9: 98944158-98944158 on Build GRCh38
rs765058784 Chr.9: 98944233-98944233 on Build GRCh38
rs200866045 Chr.9: 98944196-98944196 on Build GRCh38
rs369351245 Chr.9: 98944171-98944171 on Build GRCh38
rs754932175 Chr.9: 98944036-98944038 on Build GRCh38
rs765903313 Chr.9: 98944044-98944044 on Build GRCh38
rs749712321 Chr.9: 98944203-98944203 on Build GRCh38
rs756018296 Chr.9: 98944058-98944058 on Build GRCh38
rs777692272 Chr.9: 98944067-98944067 on Build GRCh38
rs574252630 Chr.9: 98944052-98944052 on Build GRCh38
rs575245670 Chr.9: 98944127-98944127 on Build GRCh38
rs760413730 Chr.9: 98944223-98944223 on Build GRCh38
rs149989981 Chr.9: 98944237-98944237 on Build GRCh38
rs536828755 Chr.9: 98944053-98944053 on Build GRCh38
rs182109129 Chr.9: 98944134-98944134 on Build GRCh38
rs764015031 Chr.9: 98944149-98944149 on Build GRCh38
rs758301996 Chr.9: 98944163-98944164 on Build GRCh38
rs774909394 Chr.9: 98944126-98944126 on Build GRCh38
rs775028238 Chr.9: 98944219-98944219 on Build GRCh38
rs763399815 Chr.9: 98944042-98944042 on Build GRCh38
rs748380307 Chr.9: 98944103-98944103 on Build GRCh38
rs954312763 Chr.9: 98944078-98944078 on Build GRCh38
rs771281346 Chr.9: 98944210-98944210 on Build GRCh38
rs749263521 Chr.9: 98944076-98944076 on Build GRCh38
rs752580822 Chr.9: 98944105-98944105 on Build GRCh38
rs760069779 Chr.9: 98944215-98944215 on Build GRCh38
rs776757383 Chr.9: 98944100-98944100 on Build GRCh38
rs752346135 Chr.9: 98944144-98944144 on Build GRCh38

View Full Product Details