Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43773121-43773382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL3
Entrez Gene ID
80237
Gene Name
elongation factor for RNA polymerase II 3
Gene Aliases
-
UniGene
Hs.706346
Gene Chromosome Location
Chr.15: 43772600-43777304 on Build GRCh38
Transcripts
NM_025165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1373078 c.1111C>T p.R371C Chr.15: 43773199-43773199 on Build GRCh38
1373077 c.1116C>T p.R372R Chr.15: 43773194-43773194 on Build GRCh38
3744617 c.1187G>A p.G396D Chr.15: 43773123-43773123 on Build GRCh38
7413966 c.1178A>C p.K393T Chr.15: 43773132-43773132 on Build GRCh38
4680783 c.1109_1110insA p.R371fs*4 Chr.15: 43773200-43773201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766591422 Chr.15: 43773198-43773198 on Build GRCh38
rs879362413 Chr.15: 43773157-43773157 on Build GRCh38
rs117693451 Chr.15: 43773339-43773339 on Build GRCh38
rs746794951 Chr.15: 43773248-43773248 on Build GRCh38
rs939557577 Chr.15: 43773156-43773156 on Build GRCh38
rs750854788 Chr.15: 43773190-43773190 on Build GRCh38
rs777648508 Chr.15: 43773329-43773329 on Build GRCh38
rs751833360 Chr.15: 43773199-43773199 on Build GRCh38
rs1014497138 Chr.15: 43773300-43773300 on Build GRCh38
rs369350357 Chr.15: 43773284-43773284 on Build GRCh38
rs761344431 Chr.15: 43773267-43773267 on Build GRCh38
rs776233149 Chr.15: 43773263-43773263 on Build GRCh38
rs752922378 Chr.15: 43773313-43773313 on Build GRCh38
rs955714389 Chr.15: 43773348-43773348 on Build GRCh38
rs765811648 Chr.15: 43773153-43773153 on Build GRCh38
rs756112259 Chr.15: 43773221-43773221 on Build GRCh38
rs908211837 Chr.15: 43773209-43773248 on Build GRCh38
rs1004195369 Chr.15: 43773286-43773286 on Build GRCh38
rs200960192 Chr.15: 43773310-43773310 on Build GRCh38
rs202108980 Chr.15: 43773297-43773297 on Build GRCh38
rs374478061 Chr.15: 43773282-43773282 on Build GRCh38
rs758812825 Chr.15: 43773196-43773196 on Build GRCh38
rs753948777 Chr.15: 43773330-43773330 on Build GRCh38
rs746845830 Chr.15: 43773343-43773343 on Build GRCh38
rs745735240 Chr.15: 43773228-43773228 on Build GRCh38
rs201925715 Chr.15: 43773220-43773220 on Build GRCh38
rs771905866 Chr.15: 43773239-43773239 on Build GRCh38
rs764602707 Chr.15: 43773268-43773268 on Build GRCh38
rs755314954 Chr.15: 43773209-43773209 on Build GRCh38
rs758327296 Chr.15: 43773334-43773334 on Build GRCh38
rs1016026342 Chr.15: 43773291-43773291 on Build GRCh38
rs574995178 Chr.15: 43773273-43773273 on Build GRCh38
rs763299517 Chr.15: 43773274-43773274 on Build GRCh38
rs1048440995 Chr.15: 43773283-43773283 on Build GRCh38
rs897804432 Chr.15: 43773203-43773203 on Build GRCh38
rs768345315 Chr.15: 43773258-43773258 on Build GRCh38
rs371465757 Chr.15: 43773226-43773226 on Build GRCh38
rs563469630 Chr.15: 43773308-43773308 on Build GRCh38
rs147445511 Chr.15: 43773278-43773278 on Build GRCh38
rs781546294 Chr.15: 43773214-43773214 on Build GRCh38
rs775332763 Chr.15: 43773244-43773244 on Build GRCh38
rs200102908 Chr.15: 43773326-43773326 on Build GRCh38

View Full Product Details