Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32170274-32170501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT1
Entrez Gene ID
10554
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases
1-AGPAT1, G15, LPAAT-alpha, LPAATA
UniGene
Hs.409230
Gene Chromosome Location
Chr.6: 32168206-32178111 on Build GRCh38
Transcripts
NM_006411.3 NM_032741.4 XM_005248805.3 XM_005248806.2 XM_011514234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5001061 c.467T>C p.I156T Chr.6: 32170468-32170468 on Build GRCh38
7087064 c.457G>T p.G153W Chr.6: 32170478-32170478 on Build GRCh38
4927609 c.478T>C p.S160P Chr.6: 32170457-32170457 on Build GRCh38
6596357 c.451C>T p.R151C Chr.6: 32170484-32170484 on Build GRCh38
6596362 c.455C>T p.T152M Chr.6: 32170480-32170480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890809433 Chr.6: 32170357-32170357 on Build GRCh38
rs982222711 Chr.6: 32170323-32170323 on Build GRCh38
rs962400353 Chr.6: 32170430-32170430 on Build GRCh38
rs926428611 Chr.6: 32170354-32170354 on Build GRCh38
rs997138049 Chr.6: 32170329-32170329 on Build GRCh38
rs763726315 Chr.6: 32170480-32170480 on Build GRCh38
rs780102338 Chr.6: 32170367-32170367 on Build GRCh38
rs753269798 Chr.6: 32170477-32170477 on Build GRCh38
rs762484753 Chr.6: 32170479-32170479 on Build GRCh38
rs141411867 Chr.6: 32170449-32170449 on Build GRCh38
rs1015638644 Chr.6: 32170303-32170303 on Build GRCh38
rs760921280 Chr.6: 32170411-32170411 on Build GRCh38
rs971187048 Chr.6: 32170313-32170313 on Build GRCh38
rs748217549 Chr.6: 32170382-32170382 on Build GRCh38
rs771280382 Chr.6: 32170434-32170434 on Build GRCh38
rs989294584 Chr.6: 32170383-32170383 on Build GRCh38
rs915154742 Chr.6: 32170431-32170431 on Build GRCh38
rs995632838 Chr.6: 32170475-32170475 on Build GRCh38
rs937844708 Chr.6: 32170372-32170372 on Build GRCh38
rs773358832 Chr.6: 32170404-32170404 on Build GRCh38
rs1003877948 Chr.6: 32170361-32170362 on Build GRCh38
rs1042001103 Chr.6: 32170454-32170454 on Build GRCh38
rs772156441 Chr.6: 32170400-32170400 on Build GRCh38
rs552578522 Chr.6: 32170446-32170446 on Build GRCh38
rs765658820 Chr.6: 32170465-32170465 on Build GRCh38

View Full Product Details