Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35780791-35781052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP33
Entrez Gene ID
115703
Gene Name
Rho GTPase activating protein 33
Gene Aliases
NOMA-GAP, SNX26, TCGAP
UniGene
Hs.515364
Gene Chromosome Location
Chr.19: 35775515-35788822 on Build GRCh38
Transcripts
NM_001172630.1 NM_052948.3 XM_005258488.1 XM_006722999.1 XM_011526416.1 XM_011526417.1 XM_011526418.1 XM_011526419.1 XM_011526420.1 XM_011526421.2 XM_017026238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
995224 c.902G>T p.R301L Chr.19: 35780992-35780992 on Build GRCh38
6603488 c.815C>G p.S272W Chr.19: 35780802-35780802 on Build GRCh38
995225 c.954C>T p.G318G Chr.19: 35781044-35781044 on Build GRCh38
6603494 c.830-9delC p.? Chr.19: 35780911-35780911 on Build GRCh38
7128332 c.414G>A p.L138L Chr.19: 35780809-35780809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756703247 Chr.19: 35780898-35780898 on Build GRCh38
rs755156294 Chr.19: 35780907-35780907 on Build GRCh38
rs775198793 Chr.19: 35781004-35781004 on Build GRCh38
rs200661379 Chr.19: 35780812-35780812 on Build GRCh38
rs377549144 Chr.19: 35780871-35780871 on Build GRCh38
rs377724119 Chr.19: 35780979-35780979 on Build GRCh38
rs769424531 Chr.19: 35780998-35780998 on Build GRCh38
rs773032356 Chr.19: 35780844-35780844 on Build GRCh38
rs200874165 Chr.19: 35780993-35780993 on Build GRCh38
rs201824798 Chr.19: 35780985-35780985 on Build GRCh38
rs755068272 Chr.19: 35780971-35780971 on Build GRCh38
rs773677089 Chr.19: 35780961-35780961 on Build GRCh38
rs145457173 Chr.19: 35780929-35780929 on Build GRCh38
rs754929455 Chr.19: 35780897-35780897 on Build GRCh38
rs777687680 Chr.19: 35780920-35780920 on Build GRCh38
rs771272256 Chr.19: 35780992-35780992 on Build GRCh38
rs377024802 Chr.19: 35780878-35780878 on Build GRCh38
rs768397311 Chr.19: 35781016-35781016 on Build GRCh38
rs768000979 Chr.19: 35780884-35780884 on Build GRCh38
rs375841616 Chr.19: 35780832-35780832 on Build GRCh38
rs746859561 Chr.19: 35780986-35780986 on Build GRCh38
rs761057554 Chr.19: 35780963-35780963 on Build GRCh38
rs750891358 Chr.19: 35780887-35780887 on Build GRCh38
rs766943681 Chr.19: 35780967-35780967 on Build GRCh38
rs770339837 Chr.19: 35780854-35780854 on Build GRCh38
rs758613542 Chr.19: 35780987-35780988 on Build GRCh38
rs762762008 Chr.19: 35780952-35780952 on Build GRCh38
rs370215328 Chr.19: 35780834-35780834 on Build GRCh38
rs973867667 Chr.19: 35780877-35780877 on Build GRCh38
rs756153377 Chr.19: 35780816-35780816 on Build GRCh38
rs753962455 Chr.19: 35780905-35780905 on Build GRCh38
rs778620090 Chr.19: 35780900-35780901 on Build GRCh38
rs142581391 Chr.19: 35780935-35780935 on Build GRCh38
rs373692589 Chr.19: 35780970-35780970 on Build GRCh38
rs946980940 Chr.19: 35781015-35781015 on Build GRCh38
rs539159889 Chr.19: 35780977-35780977 on Build GRCh38
rs747885619 Chr.19: 35780911-35780911 on Build GRCh38
rs973734900 Chr.19: 35780914-35780914 on Build GRCh38
rs780247702 Chr.19: 35780821-35780821 on Build GRCh38
rs776261024 Chr.19: 35780857-35780857 on Build GRCh38
rs765372672 Chr.19: 35780976-35780976 on Build GRCh38
rs757101169 Chr.19: 35780991-35780991 on Build GRCh38
rs920811017 Chr.19: 35780927-35780927 on Build GRCh38
rs761126504 Chr.19: 35781020-35781020 on Build GRCh38
rs779132135 Chr.19: 35780910-35780910 on Build GRCh38
rs199888840 Chr.19: 35780846-35780846 on Build GRCh38
rs766349583 Chr.19: 35780904-35780904 on Build GRCh38
rs369417707 Chr.19: 35780827-35780827 on Build GRCh38
rs200027656 Chr.19: 35781035-35781035 on Build GRCh38
rs537843025 Chr.19: 35780831-35780831 on Build GRCh38
rs766742660 Chr.19: 35781029-35781029 on Build GRCh38
rs748927878 Chr.19: 35781014-35781014 on Build GRCh38
rs771952135 Chr.19: 35780836-35780836 on Build GRCh38
rs571665836 Chr.19: 35780908-35780908 on Build GRCh38
rs767761630 Chr.19: 35780959-35780959 on Build GRCh38
rs868230783 Chr.19: 35781003-35781003 on Build GRCh38
rs752907607 Chr.19: 35780978-35780978 on Build GRCh38
rs758128795 Chr.19: 35780917-35780917 on Build GRCh38
rs745401492 Chr.19: 35780943-35780943 on Build GRCh38
rs201001024 Chr.19: 35780948-35780948 on Build GRCh38
rs746903901 Chr.19: 35780928-35780928 on Build GRCh38
rs369391023 Chr.19: 35780882-35780882 on Build GRCh38
rs749459839 Chr.19: 35780823-35780823 on Build GRCh38

View Full Product Details