Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11546639-11546876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNN1
Entrez Gene ID
1264
Gene Name
calponin 1
Gene Aliases
HEL-S-14, SMCC, Sm-Calp
UniGene
Hs.465929
Gene Chromosome Location
Chr.19: 11538717-11550323 on Build GRCh38
Transcripts
NM_001299.5 NM_001308341.1 NM_001308342.1 XM_005259741.2 XM_017026289.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
438568 c.276C>G p.I92M Chr.19: 11546855-11546855 on Build GRCh38
6149667 c.189C>T p.F63F Chr.19: 11546678-11546678 on Build GRCh38
6317567 c.209G>T p.G70V Chr.19: 11546698-11546698 on Build GRCh38
7207742 c.264C>T p.I88I Chr.19: 11546843-11546843 on Build GRCh38
6631261 c.214G>A p.V72M Chr.19: 11546703-11546703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772757556 Chr.19: 11546779-11546780 on Build GRCh38
rs756193455 Chr.19: 11546810-11546810 on Build GRCh38
rs777785936 Chr.19: 11546723-11546723 on Build GRCh38
rs139732050 Chr.19: 11546811-11546811 on Build GRCh38
rs753809316 Chr.19: 11546822-11546822 on Build GRCh38
rs763355259 Chr.19: 11546690-11546690 on Build GRCh38
rs188693039 Chr.19: 11546694-11546694 on Build GRCh38
rs770724445 Chr.19: 11546746-11546746 on Build GRCh38
rs771723134 Chr.19: 11546759-11546759 on Build GRCh38
rs756526545 Chr.19: 11546817-11546817 on Build GRCh38
rs765805836 Chr.19: 11546662-11546662 on Build GRCh38
rs960533604 Chr.19: 11546821-11546821 on Build GRCh38
rs752832081 Chr.19: 11546702-11546702 on Build GRCh38
rs760201901 Chr.19: 11546776-11546776 on Build GRCh38
rs749229635 Chr.19: 11546744-11546744 on Build GRCh38
rs147357819 Chr.19: 11546801-11546801 on Build GRCh38
rs756073519 Chr.19: 11546714-11546714 on Build GRCh38
rs767885939 Chr.19: 11546803-11546803 on Build GRCh38
rs111426320 Chr.19: 11546673-11546673 on Build GRCh38
rs768101664 Chr.19: 11546777-11546777 on Build GRCh38
rs752773185 Chr.19: 11546804-11546804 on Build GRCh38
rs745505797 Chr.19: 11546757-11546757 on Build GRCh38
rs112362401 Chr.19: 11546674-11546674 on Build GRCh38
rs778640677 Chr.19: 11546754-11546754 on Build GRCh38
rs560520388 Chr.19: 11546826-11546826 on Build GRCh38
rs368596033 Chr.19: 11546825-11546825 on Build GRCh38
rs751805904 Chr.19: 11546692-11546692 on Build GRCh38
rs369851055 Chr.19: 11546685-11546685 on Build GRCh38
rs140329825 Chr.19: 11546843-11546843 on Build GRCh38
rs781366485 Chr.19: 11546696-11546696 on Build GRCh38
rs565378467 Chr.19: 11546781-11546792 on Build GRCh38
rs774913019 Chr.19: 11546775-11546775 on Build GRCh38
rs763437651 Chr.19: 11546788-11546788 on Build GRCh38
rs750705682 Chr.19: 11546664-11546664 on Build GRCh38
rs773738334 Chr.19: 11546783-11546783 on Build GRCh38
rs867112592 Chr.19: 11546852-11546852 on Build GRCh38
rs1002927174 Chr.19: 11546778-11546778 on Build GRCh38

View Full Product Details