Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144394421-144394650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF1
Entrez Gene ID
29894
Gene Name
cleavage and polyadenylation specific factor 1
Gene Aliases
CPSF160, HSU37012, P/cl.18
UniGene
Hs.493202
Gene Chromosome Location
Chr.8: 144393231-144409400 on Build GRCh38
Transcripts
NM_013291.2 XM_006716548.2 XM_006716549.2 XM_006716550.3 XM_011516997.1 XM_011516998.1 XM_011516999.1

SNP Detail

SNP ID Chromosome Location
rs782001473 Chr.8: 144394573-144394573 on Build GRCh38
rs782176835 Chr.8: 144394593-144394593 on Build GRCh38
rs782430609 Chr.8: 144394462-144394462 on Build GRCh38
rs782330005 Chr.8: 144394617-144394617 on Build GRCh38
rs202060669 Chr.8: 144394471-144394471 on Build GRCh38
rs549098783 Chr.8: 144394612-144394612 on Build GRCh38
rs782025628 Chr.8: 144394463-144394463 on Build GRCh38
rs782525119 Chr.8: 144394537-144394537 on Build GRCh38
rs562038809 Chr.8: 144394481-144394481 on Build GRCh38
rs782672670 Chr.8: 144394607-144394607 on Build GRCh38
rs782700712 Chr.8: 144394493-144394493 on Build GRCh38
rs782138916 Chr.8: 144394467-144394467 on Build GRCh38
rs782581631 Chr.8: 144394592-144394592 on Build GRCh38
rs570774923 Chr.8: 144394520-144394520 on Build GRCh38
rs770523342 Chr.8: 144394627-144394627 on Build GRCh38
rs200325585 Chr.8: 144394584-144394584 on Build GRCh38
rs371378352 Chr.8: 144394500-144394500 on Build GRCh38
rs781963326 Chr.8: 144394613-144394613 on Build GRCh38
rs782150090 Chr.8: 144394505-144394505 on Build GRCh38
rs782725466 Chr.8: 144394525-144394525 on Build GRCh38
rs376203383 Chr.8: 144394579-144394579 on Build GRCh38
rs782114542 Chr.8: 144394574-144394574 on Build GRCh38
rs781944369 Chr.8: 144394457-144394457 on Build GRCh38
rs180906707 Chr.8: 144394597-144394597 on Build GRCh38
rs368299954 Chr.8: 144394580-144394580 on Build GRCh38
rs782284221 Chr.8: 144394570-144394570 on Build GRCh38
rs567207442 Chr.8: 144394629-144394629 on Build GRCh38
rs372025864 Chr.8: 144394586-144394586 on Build GRCh38
rs782257147 Chr.8: 144394610-144394610 on Build GRCh38
rs782215918 Chr.8: 144394615-144394615 on Build GRCh38
rs528682775 Chr.8: 144394623-144394623 on Build GRCh38
rs782320837 Chr.8: 144394564-144394564 on Build GRCh38
rs782639339 Chr.8: 144394444-144394444 on Build GRCh38
rs371593272 Chr.8: 144394536-144394536 on Build GRCh38
rs200863382 Chr.8: 144394572-144394572 on Build GRCh38
rs782537658 Chr.8: 144394598-144394598 on Build GRCh38
rs373469158 Chr.8: 144394624-144394624 on Build GRCh38
rs782030221 Chr.8: 144394499-144394499 on Build GRCh38
rs782399147 Chr.8: 144394559-144394560 on Build GRCh38
rs144019140 Chr.8: 144394511-144394511 on Build GRCh38
rs188385554 Chr.8: 144394585-144394585 on Build GRCh38
rs782226248 Chr.8: 144394445-144394445 on Build GRCh38
rs782009042 Chr.8: 144394626-144394626 on Build GRCh38
rs375387970 Chr.8: 144394590-144394590 on Build GRCh38
rs4451340 Chr.8: 144394566-144394566 on Build GRCh38
rs146450154 Chr.8: 144394547-144394547 on Build GRCh38
rs781912333 Chr.8: 144394569-144394569 on Build GRCh38
rs569028050 Chr.8: 144394460-144394460 on Build GRCh38
rs184042791 Chr.8: 144394448-144394448 on Build GRCh38
rs543962740 Chr.8: 144394529-144394529 on Build GRCh38
rs781908675 Chr.8: 144394583-144394583 on Build GRCh38
rs782708748 Chr.8: 144394581-144394581 on Build GRCh38
rs782211136 Chr.8: 144394563-144394563 on Build GRCh38
rs782745137 Chr.8: 144394576-144394576 on Build GRCh38
rs781930752 Chr.8: 144394569-144394570 on Build GRCh38
rs782640306 Chr.8: 144394561-144394561 on Build GRCh38
rs139096996 Chr.8: 144394472-144394472 on Build GRCh38
rs782343028 Chr.8: 144394447-144394447 on Build GRCh38

View Full Product Details