Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 175113895-175114124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF2
Entrez Gene ID
1386
Gene Name
activating transcription factor 2
Gene Aliases
CRE-BP1, CREB-2, CREB2, HB16, TREB7
UniGene
Hs.592510
Gene Chromosome Location
Chr.2: 175072250-175168206 on Build GRCh38
Transcripts
NM_001256090.1 NM_001256091.1 NM_001256092.1 NM_001256093.1 NM_001880.3 NR_045768.1 NR_045769.1 NR_045770.1 NR_045771.1 NR_045772.1 NR_045773.1 NR_045774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4968267 c.741+1G>C p.? Chr.2: 175113993-175113993 on Build GRCh38
717676 c.688G>T p.A230S Chr.2: 175114047-175114047 on Build GRCh38
6606511 c.641C>A p.P214Q Chr.2: 175114094-175114094 on Build GRCh38
1009809 c.705T>C p.I235I Chr.2: 175114030-175114030 on Build GRCh38
1009808 c.730G>A p.A244T Chr.2: 175114005-175114005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114644677 Chr.2: 175113974-175113974 on Build GRCh38
rs779874682 Chr.2: 175114011-175114011 on Build GRCh38
rs376924681 Chr.2: 175114040-175114040 on Build GRCh38
rs945250604 Chr.2: 175113977-175113977 on Build GRCh38
rs200549535 Chr.2: 175114068-175114068 on Build GRCh38
rs778993699 Chr.2: 175113998-175113998 on Build GRCh38
rs566991652 Chr.2: 175113966-175113966 on Build GRCh38
rs867049575 Chr.2: 175114038-175114038 on Build GRCh38
rs758289753 Chr.2: 175114003-175114003 on Build GRCh38
rs770962595 Chr.2: 175114053-175114053 on Build GRCh38
rs755763385 Chr.2: 175113953-175113953 on Build GRCh38
rs369150975 Chr.2: 175113963-175113963 on Build GRCh38
rs773250879 Chr.2: 175114032-175114032 on Build GRCh38
rs1042682231 Chr.2: 175113986-175113986 on Build GRCh38
rs745760497 Chr.2: 175114001-175114001 on Build GRCh38
rs748387664 Chr.2: 175114012-175114012 on Build GRCh38
rs370376829 Chr.2: 175113944-175113944 on Build GRCh38
rs1007687506 Chr.2: 175114039-175114039 on Build GRCh38
rs770045341 Chr.2: 175114016-175114016 on Build GRCh38

View Full Product Details