Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31459813-31460040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC5
Entrez Gene ID
79798
Gene Name
armadillo repeat containing 5
Gene Aliases
AIMAH2
UniGene
Hs.732945
Gene Chromosome Location
Chr.16: 31458273-31467167 on Build GRCh38
Transcripts
NM_001105247.1 NM_001288767.1 NM_001301820.1 NM_024742.2 XM_006721091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6563155 c.399C>G p.L133L Chr.16: 31459923-31459923 on Build GRCh38
3988453 c.730G>T p.V244L Chr.16: 31459969-31459969 on Build GRCh38
7226599 c.441C>A p.T147T Chr.16: 31459965-31459965 on Build GRCh38
3509241 c.617C>T p.P206L Chr.16: 31459856-31459856 on Build GRCh38
4489470 c.632C>T p.S211L Chr.16: 31459871-31459871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976862231 Chr.16: 31459854-31459854 on Build GRCh38
rs538649451 Chr.16: 31459866-31459868 on Build GRCh38
rs140265866 Chr.16: 31459914-31459914 on Build GRCh38
rs759972257 Chr.16: 31459883-31459883 on Build GRCh38
rs752908182 Chr.16: 31459898-31459898 on Build GRCh38
rs1050026325 Chr.16: 31459864-31459864 on Build GRCh38
rs184376263 Chr.16: 31459866-31459866 on Build GRCh38
rs771845578 Chr.16: 31459931-31459931 on Build GRCh38
rs1015882305 Chr.16: 31460008-31460008 on Build GRCh38
rs756248578 Chr.16: 31459853-31459853 on Build GRCh38
rs373860441 Chr.16: 31459861-31459861 on Build GRCh38
rs760080420 Chr.16: 31460011-31460011 on Build GRCh38
rs777779636 Chr.16: 31459858-31459858 on Build GRCh38
rs114930262 Chr.16: 31459990-31459990 on Build GRCh38
rs983210811 Chr.16: 31459843-31459843 on Build GRCh38
rs776051274 Chr.16: 31459938-31459938 on Build GRCh38
rs747668287 Chr.16: 31460009-31460009 on Build GRCh38
rs1011892852 Chr.16: 31459989-31459989 on Build GRCh38
rs766609669 Chr.16: 31459846-31459846 on Build GRCh38
rs757191392 Chr.16: 31459920-31459920 on Build GRCh38
rs753906161 Chr.16: 31459964-31459964 on Build GRCh38
rs759992443 Chr.16: 31459953-31459953 on Build GRCh38
rs771753877 Chr.16: 31459868-31459868 on Build GRCh38
rs780891347 Chr.16: 31459996-31459996 on Build GRCh38
rs770791789 Chr.16: 31459863-31459863 on Build GRCh38
rs778882433 Chr.16: 31459926-31459926 on Build GRCh38
rs779697865 Chr.16: 31459932-31459932 on Build GRCh38
rs750885418 Chr.16: 31459839-31459839 on Build GRCh38
rs374754128 Chr.16: 31459963-31459963 on Build GRCh38
rs866327048 Chr.16: 31459973-31459973 on Build GRCh38
rs777675096 Chr.16: 31459916-31459916 on Build GRCh38
rs763317033 Chr.16: 31459875-31459875 on Build GRCh38
rs750307035 Chr.16: 31459979-31459979 on Build GRCh38
rs758217532 Chr.16: 31459985-31459985 on Build GRCh38
rs775123648 Chr.16: 31459869-31459869 on Build GRCh38
rs200289596 Chr.16: 31460010-31460010 on Build GRCh38
rs200910451 Chr.16: 31459956-31459956 on Build GRCh38
rs267604526 Chr.16: 31459993-31459993 on Build GRCh38
rs1002622442 Chr.16: 31459991-31459991 on Build GRCh38
rs946954430 Chr.16: 31459887-31459887 on Build GRCh38
rs765844255 Chr.16: 31459836-31459836 on Build GRCh38
rs771406733 Chr.16: 31459944-31459944 on Build GRCh38
rs758790821 Chr.16: 31459842-31459842 on Build GRCh38
rs766884244 Chr.16: 31459876-31459876 on Build GRCh38
rs199693319 Chr.16: 31459867-31459867 on Build GRCh38
rs774984127 Chr.16: 31459951-31459951 on Build GRCh38
rs773795693 Chr.16: 31459872-31459872 on Build GRCh38
rs201768837 Chr.16: 31459852-31459852 on Build GRCh38
rs548290271 Chr.16: 31459850-31459850 on Build GRCh38
rs768230478 Chr.16: 31459994-31459994 on Build GRCh38
rs761353040 Chr.16: 31459941-31459941 on Build GRCh38
rs760240565 Chr.16: 31459870-31459870 on Build GRCh38
rs200926753 Chr.16: 31459965-31459965 on Build GRCh38
rs376806624 Chr.16: 31460012-31460012 on Build GRCh38
rs201280100 Chr.16: 31459962-31459962 on Build GRCh38
rs767908538 Chr.16: 31459897-31459897 on Build GRCh38
rs538571383 Chr.16: 31459904-31459904 on Build GRCh38
rs374896159 Chr.16: 31459878-31459878 on Build GRCh38
rs574798073 Chr.16: 31459955-31459955 on Build GRCh38
rs778883640 Chr.16: 31459978-31459978 on Build GRCh38
rs760150629 Chr.16: 31459832-31459832 on Build GRCh38
rs745732463 Chr.16: 31459930-31459930 on Build GRCh38
rs932874365 Chr.16: 31459857-31459857 on Build GRCh38
rs369520261 Chr.16: 31460014-31460014 on Build GRCh38
rs776047628 Chr.16: 31460013-31460013 on Build GRCh38
rs571045241 Chr.16: 31459871-31459871 on Build GRCh38
rs753838016 Chr.16: 31459917-31459917 on Build GRCh38

View Full Product Details