Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36611991-36612264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf24
Entrez Gene ID
55969
Gene Name
chromosome 20 open reading frame 24
Gene Aliases
PNAS-11, RIP5
UniGene
Hs.584985
Gene Chromosome Location
Chr.20: 36605734-36612557 on Build GRCh38
Transcripts
NM_001199534.1 NM_018840.4 NM_199483.2 NR_026562.3 NM_001199535.1

Gene Symbol
TGIF2-C20orf24
Entrez Gene ID
100527943
Gene Name
TGIF2-C20orf24 readthrough
Gene Aliases
-
UniGene
Hs.584985
Gene Chromosome Location
Chr.20: 36574554-36612557 on Build GRCh38
Transcripts
NM_001199534.1 NM_018840.4 NM_199483.2 NR_026562.3 NM_001199535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4485127 c.289C>A p.P97T Chr.20: 36612080-36612080 on Build GRCh38
723860 c.356G>T p.W119L Chr.20: 36612017-36612017 on Build GRCh38
5462954 c.315A>G p.T105T Chr.20: 36612106-36612106 on Build GRCh38
3939322 c.219-2A>G p.? Chr.20: 36612008-36612008 on Build GRCh38
3545877 c.286T>C p.Y96H Chr.20: 36612077-36612077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754188455 Chr.20: 36612197-36612197 on Build GRCh38
rs774446475 Chr.20: 36612165-36612165 on Build GRCh38
rs775836719 Chr.20: 36612018-36612019 on Build GRCh38
rs200804664 Chr.20: 36612110-36612110 on Build GRCh38
rs776267934 Chr.20: 36612058-36612058 on Build GRCh38
rs753030412 Chr.20: 36612094-36612094 on Build GRCh38
rs190742083 Chr.20: 36612122-36612122 on Build GRCh38
rs140513057 Chr.20: 36612137-36612137 on Build GRCh38
rs369227155 Chr.20: 36612060-36612060 on Build GRCh38
rs1010758693 Chr.20: 36612133-36612133 on Build GRCh38
rs747875227 Chr.20: 36612153-36612153 on Build GRCh38
rs746541430 Chr.20: 36612055-36612055 on Build GRCh38
rs202088044 Chr.20: 36612157-36612157 on Build GRCh38
rs555687131 Chr.20: 36612205-36612205 on Build GRCh38
rs764118873 Chr.20: 36612227-36612228 on Build GRCh38
rs765136763 Chr.20: 36612064-36612064 on Build GRCh38
rs924073294 Chr.20: 36612096-36612096 on Build GRCh38
rs751651327 Chr.20: 36612081-36612081 on Build GRCh38
rs6028360 Chr.20: 36612210-36612210 on Build GRCh38
rs750986212 Chr.20: 36612211-36612211 on Build GRCh38
rs138333574 Chr.20: 36612159-36612159 on Build GRCh38
rs771447581 Chr.20: 36612050-36612050 on Build GRCh38
rs766251711 Chr.20: 36612078-36612078 on Build GRCh38
rs775027581 Chr.20: 36612054-36612054 on Build GRCh38
rs372092965 Chr.20: 36612083-36612083 on Build GRCh38
rs375889787 Chr.20: 36612150-36612150 on Build GRCh38
rs763131205 Chr.20: 36612116-36612117 on Build GRCh38
rs551295814 Chr.20: 36612099-36612099 on Build GRCh38
rs772993204 Chr.20: 36612066-36612066 on Build GRCh38
rs143917410 Chr.20: 36612103-36612103 on Build GRCh38
rs1018418843 Chr.20: 36612135-36612135 on Build GRCh38
rs138244240 Chr.20: 36612020-36612020 on Build GRCh38
rs746447467 Chr.20: 36612124-36612124 on Build GRCh38
rs566354083 Chr.20: 36612123-36612123 on Build GRCh38
rs150570188 Chr.20: 36612208-36612208 on Build GRCh38
rs534176887 Chr.20: 36612160-36612160 on Build GRCh38
rs141883588 Chr.20: 36612175-36612175 on Build GRCh38
rs138992970 Chr.20: 36612090-36612090 on Build GRCh38
rs2232911 Chr.20: 36612154-36612154 on Build GRCh38
rs376043012 Chr.20: 36612176-36612176 on Build GRCh38
rs762996706 Chr.20: 36612076-36612076 on Build GRCh38
rs200128148 Chr.20: 36612173-36612173 on Build GRCh38
rs145156759 Chr.20: 36612056-36612056 on Build GRCh38
rs542504187 Chr.20: 36612116-36612116 on Build GRCh38
rs367602721 Chr.20: 36612172-36612172 on Build GRCh38
rs935268879 Chr.20: 36612105-36612105 on Build GRCh38

View Full Product Details